Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103119971-103120216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPE
Entrez Gene ID
1062
Gene Name
centromere protein E
Gene Aliases
CENP-E, KIF10, MCPH13, PPP1R61
UniGene
Hs.75573
Gene Chromosome Location
Chr.4: 103105806-103198409 on Build GRCh38
Transcripts
NM_001286734.1 NM_001813.2 XM_011531544.2 XM_011531545.2 XM_011531546.2 XM_011531547.2 XM_011531548.2 XM_011531549.2 XM_017007659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376457 c.7318C>A p.Q2440K Chr.4: 103120159-103120159 on Build GRCh38
6376458 c.7207C>A p.Q2403K Chr.4: 103120159-103120159 on Build GRCh38
6098781 c.7176T>A p.N2392K Chr.4: 103120190-103120190 on Build GRCh38
6098780 c.7287T>A p.N2429K Chr.4: 103120190-103120190 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779966149 Chr.4: 103120098-103120098 on Build GRCh38
rs957596275 Chr.4: 103120068-103120068 on Build GRCh38
rs550484015 Chr.4: 103120101-103120102 on Build GRCh38
rs990198740 Chr.4: 103120085-103120085 on Build GRCh38
rs142518565 Chr.4: 103120183-103120183 on Build GRCh38
rs768415435 Chr.4: 103120107-103120107 on Build GRCh38
rs774078104 Chr.4: 103120117-103120117 on Build GRCh38
rs115120218 Chr.4: 103120149-103120149 on Build GRCh38
rs945297310 Chr.4: 103120121-103120121 on Build GRCh38
rs765636963 Chr.4: 103120159-103120159 on Build GRCh38
rs750973281 Chr.4: 103120129-103120129 on Build GRCh38
rs775817203 Chr.4: 103120166-103120166 on Build GRCh38
rs1004989900 Chr.4: 103120005-103120005 on Build GRCh38
rs760057241 Chr.4: 103120157-103120157 on Build GRCh38
rs757630463 Chr.4: 103120119-103120120 on Build GRCh38
rs764306851 Chr.4: 103120182-103120182 on Build GRCh38
rs1002659960 Chr.4: 103120078-103120078 on Build GRCh38
rs1028956979 Chr.4: 103120059-103120059 on Build GRCh38
rs1019091365 Chr.4: 103120022-103120022 on Build GRCh38
rs190881104 Chr.4: 103120036-103120036 on Build GRCh38
rs182927736 Chr.4: 103120102-103120102 on Build GRCh38
rs1056020613 Chr.4: 103120095-103120095 on Build GRCh38
rs779356833 Chr.4: 103120125-103120126 on Build GRCh38
rs573435601 Chr.4: 103120027-103120027 on Build GRCh38
rs927794104 Chr.4: 103120142-103120142 on Build GRCh38
rs2720453 Chr.4: 103120136-103120136 on Build GRCh38
rs914219191 Chr.4: 103120002-103120002 on Build GRCh38
rs915147055 Chr.4: 103120090-103120090 on Build GRCh38
rs763228807 Chr.4: 103120172-103120172 on Build GRCh38
rs529099827 Chr.4: 103120053-103120053 on Build GRCh38
rs1054067277 Chr.4: 103120000-103120000 on Build GRCh38
rs532716630 Chr.4: 103120060-103120060 on Build GRCh38
rs1026870204 Chr.4: 103120105-103120105 on Build GRCh38
rs757307598 Chr.4: 103120185-103120185 on Build GRCh38
rs771558355 Chr.4: 103120137-103120137 on Build GRCh38
rs767168339 Chr.4: 103120102-103120105 on Build GRCh38

View Full Product Details