Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80695712-80695939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD17C
Entrez Gene ID
58489
Gene Name
abhydrolase domain containing 17C
Gene Aliases
FAM108C1
UniGene
Hs.459072
Gene Chromosome Location
Chr.15: 80695311-80755621 on Build GRCh38
Transcripts
NM_021214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363001 c.309G>A p.E103E Chr.15: 80695738-80695738 on Build GRCh38
6423942 c.366C>A p.C122* Chr.15: 80695795-80695795 on Build GRCh38
5427205 c.388T>C p.S130P Chr.15: 80695817-80695817 on Build GRCh38
3794453 c.470C>T p.S157F Chr.15: 80695899-80695899 on Build GRCh38
6585220 c.330C>A p.F110L Chr.15: 80695759-80695759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773645113 Chr.15: 80695897-80695897 on Build GRCh38
rs761526321 Chr.15: 80695747-80695747 on Build GRCh38
rs552694620 Chr.15: 80695790-80695790 on Build GRCh38
rs749248155 Chr.15: 80695858-80695858 on Build GRCh38
rs747783328 Chr.15: 80695885-80695885 on Build GRCh38
rs754476870 Chr.15: 80695862-80695862 on Build GRCh38
rs780031599 Chr.15: 80695840-80695840 on Build GRCh38
rs1052579165 Chr.15: 80695801-80695801 on Build GRCh38
rs765312505 Chr.15: 80695908-80695908 on Build GRCh38
rs74342683 Chr.15: 80695811-80695811 on Build GRCh38
rs777167453 Chr.15: 80695902-80695902 on Build GRCh38
rs755990261 Chr.15: 80695827-80695827 on Build GRCh38
rs778332098 Chr.15: 80695882-80695882 on Build GRCh38
rs752941756 Chr.15: 80695789-80695789 on Build GRCh38
rs569270377 Chr.15: 80695819-80695819 on Build GRCh38
rs758642384 Chr.15: 80695805-80695805 on Build GRCh38
rs763098890 Chr.15: 80695915-80695915 on Build GRCh38
rs747388882 Chr.15: 80695898-80695898 on Build GRCh38
rs538251048 Chr.15: 80695887-80695887 on Build GRCh38
rs771491331 Chr.15: 80695899-80695899 on Build GRCh38
rs753975532 Chr.15: 80695836-80695836 on Build GRCh38
rs936818747 Chr.15: 80695765-80695765 on Build GRCh38
rs533550000 Chr.15: 80695916-80695916 on Build GRCh38
rs555731871 Chr.15: 80695756-80695756 on Build GRCh38
rs914147043 Chr.15: 80695810-80695810 on Build GRCh38
rs759659051 Chr.15: 80695768-80695768 on Build GRCh38
rs201033272 Chr.15: 80695909-80695909 on Build GRCh38
rs780928658 Chr.15: 80695817-80695817 on Build GRCh38
rs765275119 Chr.15: 80695772-80695772 on Build GRCh38
rs753801646 Chr.15: 80695762-80695762 on Build GRCh38

View Full Product Details