Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50616009-50616274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRB10
Entrez Gene ID
2887
Gene Name
growth factor receptor bound protein 10
Gene Aliases
GRB-IR, Grb-10, IRBP, MEG1, RSS
UniGene
Hs.164060
Gene Chromosome Location
Chr.7: 50590063-50793462 on Build GRCh38
Transcripts
NM_001001549.2 NM_001001550.2 NM_001001555.2 NM_005311.4 XM_011515304.1 XM_011515312.2 XM_011515316.1 XM_011515323.2 XM_017012029.1 XM_017012030.1 XM_017012031.1 XM_017012032.1 XM_017012033.1 XM_017012034.1 XM_017012035.1 XM_017012036.1 XM_017012037.1 XM_017012038.1 XM_017012039.1 XM_017012040.1 XM_017012041.1 XM_017012042.1 XM_017012043.1 XM_017012044.1 XM_017012045.1 XM_017012046.1 XM_017012047.1 XM_017012048.1 XM_017012049.1 XM_017012050.1 XM_017012051.1 XM_017012052.1 XM_017012053.1 XM_017012054.1 XM_017012055.1 XM_017012056.1 XM_017012057.1 XM_017012058.1 XM_017012059.1 XM_017012060.1 XM_017012061.1 XM_017012062.1 XM_017012063.1 XM_017012064.1 XM_017012065.1 XM_017012066.1 XM_017012067.1 XM_017012068.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7298021 c.943C>T p.R315W Chr.7: 50616251-50616251 on Build GRCh38
3265627 c.966+1G>T p.? Chr.7: 50616209-50616209 on Build GRCh38
3881655 c.935G>A p.G312D Chr.7: 50616241-50616241 on Build GRCh38
1208833 c.926G>A p.R309Q Chr.7: 50616250-50616250 on Build GRCh38
1159285 c.966+1G>C p.? Chr.7: 50616209-50616209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576871336 Chr.7: 50616049-50616049 on Build GRCh38
rs374440919 Chr.7: 50616181-50616181 on Build GRCh38
rs750610949 Chr.7: 50616153-50616153 on Build GRCh38
rs189295544 Chr.7: 50616044-50616044 on Build GRCh38
rs764158392 Chr.7: 50616117-50616117 on Build GRCh38
rs1057336805 Chr.7: 50616071-50616071 on Build GRCh38
rs1026598569 Chr.7: 50616149-50616149 on Build GRCh38
rs975168343 Chr.7: 50616199-50616199 on Build GRCh38
rs1009085768 Chr.7: 50616157-50616157 on Build GRCh38
rs199551488 Chr.7: 50616163-50616163 on Build GRCh38
rs200796219 Chr.7: 50616226-50616226 on Build GRCh38
rs745575041 Chr.7: 50616189-50616189 on Build GRCh38
rs368562566 Chr.7: 50616251-50616251 on Build GRCh38
rs899205469 Chr.7: 50616120-50616120 on Build GRCh38
rs572787303 Chr.7: 50616103-50616103 on Build GRCh38
rs924512702 Chr.7: 50616038-50616038 on Build GRCh38
rs901334880 Chr.7: 50616195-50616195 on Build GRCh38
rs760161025 Chr.7: 50616159-50616159 on Build GRCh38
rs765036374 Chr.7: 50616174-50616174 on Build GRCh38
rs539619039 Chr.7: 50616068-50616068 on Build GRCh38
rs561669666 Chr.7: 50616162-50616162 on Build GRCh38
rs536356257 Chr.7: 50616155-50616155 on Build GRCh38
rs778637273 Chr.7: 50616182-50616182 on Build GRCh38
rs755087835 Chr.7: 50616198-50616198 on Build GRCh38
rs895917003 Chr.7: 50616085-50616085 on Build GRCh38
rs768200199 Chr.7: 50616160-50616160 on Build GRCh38
rs766706818 Chr.7: 50616185-50616185 on Build GRCh38
rs541751041 Chr.7: 50616134-50616134 on Build GRCh38
rs948773079 Chr.7: 50616096-50616096 on Build GRCh38
rs894344022 Chr.7: 50616154-50616154 on Build GRCh38
rs751888247 Chr.7: 50616186-50616186 on Build GRCh38
rs192228326 Chr.7: 50616101-50616101 on Build GRCh38

View Full Product Details