Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 71138391-71138664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPHOSPH10
Entrez Gene ID
10199
Gene Name
M-phase phosphoprotein 10
Gene Aliases
CT90, MPP10, MPP10P, PPP1R106
UniGene
Hs.656208
Gene Chromosome Location
Chr.2: 71130314-71150102 on Build GRCh38
Transcripts
NM_005791.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158609 c.1118C>G p.S373C Chr.2: 71138509-71138509 on Build GRCh38
1022599 c.1234C>T p.R412W Chr.2: 71138625-71138625 on Build GRCh38
1409258 c.1106delA p.I371fs*4 Chr.2: 71138497-71138497 on Build GRCh38
1409259 c.1138_1139insA p.P383fs*26 Chr.2: 71138529-71138530 on Build GRCh38
6296149 c.1185G>T p.R395S Chr.2: 71138576-71138576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750915614 Chr.2: 71138539-71138539 on Build GRCh38
rs576247993 Chr.2: 71138534-71138534 on Build GRCh38
rs763564970 Chr.2: 71138441-71138441 on Build GRCh38
rs944811517 Chr.2: 71138567-71138567 on Build GRCh38
rs1014777466 Chr.2: 71138450-71138450 on Build GRCh38
rs748249290 Chr.2: 71138485-71138485 on Build GRCh38
rs554779004 Chr.2: 71138525-71138525 on Build GRCh38
rs770925225 Chr.2: 71138502-71138502 on Build GRCh38
rs749492129 Chr.2: 71138466-71138466 on Build GRCh38
rs754174928 Chr.2: 71138548-71138548 on Build GRCh38
rs750691063 Chr.2: 71138451-71138451 on Build GRCh38
rs935153395 Chr.2: 71138561-71138561 on Build GRCh38
rs755192046 Chr.2: 71138559-71138559 on Build GRCh38
rs912765008 Chr.2: 71138438-71138438 on Build GRCh38
rs769651496 Chr.2: 71138603-71138603 on Build GRCh38
rs893171251 Chr.2: 71138588-71138588 on Build GRCh38
rs754074281 Chr.2: 71138515-71138515 on Build GRCh38
rs916509314 Chr.2: 71138522-71138522 on Build GRCh38
rs769027954 Chr.2: 71138477-71138477 on Build GRCh38
rs745788802 Chr.2: 71138591-71138591 on Build GRCh38
rs768234906 Chr.2: 71138625-71138625 on Build GRCh38
rs772266253 Chr.2: 71138581-71138581 on Build GRCh38
rs776704638 Chr.2: 71138503-71138503 on Build GRCh38
rs756483951 Chr.2: 71138456-71138456 on Build GRCh38
rs762671700 Chr.2: 71138618-71138618 on Build GRCh38
rs954255190 Chr.2: 71138433-71138433 on Build GRCh38
rs756617224 Chr.2: 71138540-71138540 on Build GRCh38
rs185323253 Chr.2: 71138568-71138568 on Build GRCh38
rs762630116 Chr.2: 71138518-71138518 on Build GRCh38
rs547565231 Chr.2: 71138437-71138437 on Build GRCh38
rs748447456 Chr.2: 71138562-71138562 on Build GRCh38
rs775308855 Chr.2: 71138614-71138614 on Build GRCh38
rs763760770 Chr.2: 71138520-71138520 on Build GRCh38
rs372174438 Chr.2: 71138532-71138532 on Build GRCh38
rs889779343 Chr.2: 71138621-71138621 on Build GRCh38
rs961941240 Chr.2: 71138460-71138460 on Build GRCh38
rs777765007 Chr.2: 71138582-71138582 on Build GRCh38
rs369680502 Chr.2: 71138635-71138635 on Build GRCh38
rs143656591 Chr.2: 71138507-71138507 on Build GRCh38
rs180701691 Chr.2: 71138446-71138446 on Build GRCh38
rs773992268 Chr.2: 71138626-71138626 on Build GRCh38
rs757714972 Chr.2: 71138444-71138444 on Build GRCh38
rs779281832 Chr.2: 71138560-71138560 on Build GRCh38
rs902381008 Chr.2: 71138558-71138558 on Build GRCh38
rs357756 Chr.2: 71138546-71138546 on Build GRCh38
rs780326889 Chr.2: 71138463-71138463 on Build GRCh38

View Full Product Details