Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3228061-3228318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRE
Entrez Gene ID
116534
Gene Name
MAS related GPR family member E
Gene Aliases
GPR167, MGRF, MRGE
UniGene
Hs.745512
Gene Chromosome Location
Chr.11: 3227811-3232386 on Build GRCh38
Transcripts
NM_001039165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638419 c.698G>A p.G233D Chr.11: 3228099-3228099 on Build GRCh38
3943632 c.509G>C p.R170P Chr.11: 3228288-3228288 on Build GRCh38
7377548 c.502C>A p.P168T Chr.11: 3228295-3228295 on Build GRCh38
5657648 c.485C>T p.T162I Chr.11: 3228312-3228312 on Build GRCh38
926363 c.599_601delTGC p.L200delL Chr.11: 3228196-3228198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531538794 Chr.11: 3228116-3228116 on Build GRCh38
rs769444901 Chr.11: 3228200-3228200 on Build GRCh38
rs763857141 Chr.11: 3228144-3228144 on Build GRCh38
rs760152909 Chr.11: 3228226-3228226 on Build GRCh38
rs368237602 Chr.11: 3228082-3228082 on Build GRCh38
rs199937193 Chr.11: 3228261-3228261 on Build GRCh38
rs372243981 Chr.11: 3228101-3228101 on Build GRCh38
rs767150972 Chr.11: 3228217-3228217 on Build GRCh38
rs199635481 Chr.11: 3228135-3228135 on Build GRCh38
rs758817154 Chr.11: 3228252-3228252 on Build GRCh38
rs374939818 Chr.11: 3228104-3228104 on Build GRCh38
rs771099077 Chr.11: 3228112-3228112 on Build GRCh38
rs888440379 Chr.11: 3228165-3228165 on Build GRCh38
rs547922965 Chr.11: 3228205-3228205 on Build GRCh38
rs765646641 Chr.11: 3228172-3228172 on Build GRCh38
rs753100615 Chr.11: 3228248-3228248 on Build GRCh38
rs547469383 Chr.11: 3228175-3228175 on Build GRCh38
rs977852583 Chr.11: 3228196-3228196 on Build GRCh38
rs756116749 Chr.11: 3228277-3228277 on Build GRCh38
rs768287313 Chr.11: 3228199-3228199 on Build GRCh38
rs749122433 Chr.11: 3228289-3228289 on Build GRCh38
rs775268366 Chr.11: 3228191-3228191 on Build GRCh38
rs904244621 Chr.11: 3228130-3228130 on Build GRCh38
rs985698013 Chr.11: 3228106-3228106 on Build GRCh38
rs749148152 Chr.11: 3228195-3228195 on Build GRCh38
rs575648651 Chr.11: 3228089-3228089 on Build GRCh38
rs1052744069 Chr.11: 3228149-3228149 on Build GRCh38
rs76136953 Chr.11: 3228187-3228187 on Build GRCh38
rs574441864 Chr.11: 3228085-3228085 on Build GRCh38
rs765809701 Chr.11: 3228246-3228246 on Build GRCh38
rs781356453 Chr.11: 3228184-3228184 on Build GRCh38
rs369020369 Chr.11: 3228113-3228113 on Build GRCh38
rs754345598 Chr.11: 3228169-3228169 on Build GRCh38
rs761387429 Chr.11: 3228208-3228208 on Build GRCh38
rs985166450 Chr.11: 3228254-3228254 on Build GRCh38
rs1030505982 Chr.11: 3228181-3228181 on Build GRCh38
rs766954894 Chr.11: 3228163-3228163 on Build GRCh38
rs759789480 Chr.11: 3228090-3228090 on Build GRCh38
rs957686838 Chr.11: 3228224-3228224 on Build GRCh38
rs897201149 Chr.11: 3228145-3228145 on Build GRCh38
rs529703230 Chr.11: 3228203-3228203 on Build GRCh38
rs780072646 Chr.11: 3228288-3228288 on Build GRCh38
rs781554426 Chr.11: 3228273-3228273 on Build GRCh38
rs765281213 Chr.11: 3228186-3228186 on Build GRCh38
rs200595956 Chr.11: 3228100-3228100 on Build GRCh38
rs368912134 Chr.11: 3228225-3228225 on Build GRCh38
rs769718336 Chr.11: 3228126-3228126 on Build GRCh38
rs373313620 Chr.11: 3228272-3228272 on Build GRCh38
rs768501750 Chr.11: 3228298-3228298 on Build GRCh38
rs1026355366 Chr.11: 3228198-3228198 on Build GRCh38
rs747184103 Chr.11: 3228180-3228180 on Build GRCh38
rs564898039 Chr.11: 3228162-3228162 on Build GRCh38
rs753152025 Chr.11: 3228174-3228174 on Build GRCh38
rs74405355 Chr.11: 3228276-3228276 on Build GRCh38
rs764310618 Chr.11: 3228255-3228255 on Build GRCh38
rs543589951 Chr.11: 3228133-3228133 on Build GRCh38
rs762643196 Chr.11: 3228134-3228134 on Build GRCh38
rs772764506 Chr.11: 3228222-3228222 on Build GRCh38
rs372474089 Chr.11: 3228132-3228132 on Build GRCh38
rs200161606 Chr.11: 3228081-3228081 on Build GRCh38
rs4405289 Chr.11: 3228171-3228171 on Build GRCh38

View Full Product Details