Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49424650-49424895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NICN1
Entrez Gene ID
84276
Gene Name
nicolin 1
Gene Aliases
-
UniGene
Hs.191460
Gene Chromosome Location
Chr.3: 49422333-49429324 on Build GRCh38
Transcripts
NM_032316.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6726082 c.614A>T p.Y205F Chr.3: 49424861-49424861 on Build GRCh38
4791660 c.616G>T p.D206Y Chr.3: 49424859-49424859 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761890566 Chr.3: 49424830-49424830 on Build GRCh38
rs565040777 Chr.3: 49424691-49424691 on Build GRCh38
rs923189240 Chr.3: 49424728-49424728 on Build GRCh38
rs751237878 Chr.3: 49424792-49424792 on Build GRCh38
rs368835208 Chr.3: 49424797-49424797 on Build GRCh38
rs200380201 Chr.3: 49424810-49424810 on Build GRCh38
rs146812560 Chr.3: 49424782-49424782 on Build GRCh38
rs143705175 Chr.3: 49424781-49424781 on Build GRCh38
rs973378464 Chr.3: 49424751-49424751 on Build GRCh38
rs776622085 Chr.3: 49424828-49424828 on Build GRCh38
rs181793435 Chr.3: 49424705-49424705 on Build GRCh38
rs896698379 Chr.3: 49424822-49424822 on Build GRCh38
rs538598133 Chr.3: 49424812-49424812 on Build GRCh38
rs189963854 Chr.3: 49424860-49424860 on Build GRCh38
rs752261821 Chr.3: 49424799-49424799 on Build GRCh38
rs756993654 Chr.3: 49424674-49424674 on Build GRCh38
rs186097465 Chr.3: 49424721-49424721 on Build GRCh38
rs866879381 Chr.3: 49424815-49424815 on Build GRCh38
rs201729332 Chr.3: 49424832-49424832 on Build GRCh38
rs745604523 Chr.3: 49424839-49424839 on Build GRCh38
rs773175517 Chr.3: 49424836-49424836 on Build GRCh38
rs757648414 Chr.3: 49424807-49424807 on Build GRCh38
rs759200754 Chr.3: 49424848-49424848 on Build GRCh38
rs780330721 Chr.3: 49424814-49424814 on Build GRCh38
rs139254977 Chr.3: 49424690-49424690 on Build GRCh38
rs754702527 Chr.3: 49424793-49424793 on Build GRCh38
rs751434221 Chr.3: 49424843-49424843 on Build GRCh38
rs369844327 Chr.3: 49424811-49424811 on Build GRCh38
rs767292718 Chr.3: 49424855-49424855 on Build GRCh38
rs367980201 Chr.3: 49424842-49424842 on Build GRCh38
rs199800701 Chr.3: 49424818-49424818 on Build GRCh38
rs768895499 Chr.3: 49424824-49424824 on Build GRCh38
rs950436877 Chr.3: 49424758-49424758 on Build GRCh38

View Full Product Details