Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218668032-218668301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF25
Entrez Gene ID
64320
Gene Name
ring finger protein 25
Gene Aliases
AO7
UniGene
Hs.471403
Gene Chromosome Location
Chr.2: 218663864-218672058 on Build GRCh38
Transcripts
NM_022453.2 XM_017004695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110711 c.269C>T p.S90L Chr.2: 218668097-218668097 on Build GRCh38
4722176 c.251G>A p.R84Q Chr.2: 218668115-218668115 on Build GRCh38
6767868 c.260G>A p.R87Q Chr.2: 218668106-218668106 on Build GRCh38
3695270 c.287+1G>A p.? Chr.2: 218668078-218668078 on Build GRCh38
1223992 c.268T>A p.S90T Chr.2: 218668098-218668098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951188488 Chr.2: 218668233-218668233 on Build GRCh38
rs747565310 Chr.2: 218668241-218668241 on Build GRCh38
rs753508264 Chr.2: 218668191-218668191 on Build GRCh38
rs746079121 Chr.2: 218668250-218668250 on Build GRCh38
rs66837497 Chr.2: 218668182-218668182 on Build GRCh38
rs775583150 Chr.2: 218668270-218668270 on Build GRCh38
rs754743322 Chr.2: 218668193-218668193 on Build GRCh38
rs571175419 Chr.2: 218668059-218668059 on Build GRCh38
rs775699312 Chr.2: 218668164-218668164 on Build GRCh38
rs771255147 Chr.2: 218668242-218668242 on Build GRCh38
rs771308052 Chr.2: 218668126-218668126 on Build GRCh38
rs772692453 Chr.2: 218668079-218668079 on Build GRCh38
rs781606543 Chr.2: 218668246-218668246 on Build GRCh38
rs200063493 Chr.2: 218668138-218668138 on Build GRCh38
rs761176212 Chr.2: 218668162-218668164 on Build GRCh38
rs773605182 Chr.2: 218668107-218668107 on Build GRCh38
rs386655241 Chr.2: 218668180-218668182 on Build GRCh38
rs891342238 Chr.2: 218668110-218668110 on Build GRCh38
rs377175164 Chr.2: 218668152-218668152 on Build GRCh38
rs999688311 Chr.2: 218668061-218668061 on Build GRCh38
rs373755582 Chr.2: 218668218-218668218 on Build GRCh38
rs757850837 Chr.2: 218668211-218668211 on Build GRCh38
rs936162688 Chr.2: 218668148-218668148 on Build GRCh38
rs532136833 Chr.2: 218668123-218668123 on Build GRCh38
rs778272851 Chr.2: 218668203-218668203 on Build GRCh38
rs112484473 Chr.2: 218668181-218668181 on Build GRCh38
rs764679568 Chr.2: 218668225-218668225 on Build GRCh38
rs115769773 Chr.2: 218668252-218668252 on Build GRCh38
rs890114134 Chr.2: 218668279-218668279 on Build GRCh38
rs2556385 Chr.2: 218668182-218668182 on Build GRCh38
rs748720228 Chr.2: 218668243-218668243 on Build GRCh38
rs751752803 Chr.2: 218668130-218668130 on Build GRCh38
rs778294037 Chr.2: 218668213-218668213 on Build GRCh38
rs2556384 Chr.2: 218668180-218668180 on Build GRCh38
rs776812226 Chr.2: 218668135-218668135 on Build GRCh38
rs926231142 Chr.2: 218668118-218668118 on Build GRCh38
rs766323653 Chr.2: 218668185-218668185 on Build GRCh38
rs376201176 Chr.2: 218668204-218668204 on Build GRCh38
rs776294554 Chr.2: 218668226-218668226 on Build GRCh38

View Full Product Details