Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35475651-35475876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEAD3
Entrez Gene ID
7005
Gene Name
TEA domain transcription factor 3
Gene Aliases
DTEF-1, ETFR-1, TEAD-3, TEAD5, TEF-5, TEF5
UniGene
Hs.485205
Gene Chromosome Location
Chr.6: 35473597-35497084 on Build GRCh38
Transcripts
NM_003214.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4161108 c.1017C>A p.D339E Chr.6: 35475701-35475701 on Build GRCh38
4161107 c.906C>A p.D302E Chr.6: 35475701-35475701 on Build GRCh38
3075517 c.1048G>A p.A350T Chr.6: 35475670-35475670 on Build GRCh38
3075516 c.937G>A p.A313T Chr.6: 35475670-35475670 on Build GRCh38
3873450 c.1014C>T p.A338A Chr.6: 35475704-35475704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756235804 Chr.6: 35475720-35475721 on Build GRCh38
rs770733961 Chr.6: 35475719-35475719 on Build GRCh38
rs369501808 Chr.6: 35475689-35475689 on Build GRCh38
rs926263772 Chr.6: 35475722-35475722 on Build GRCh38
rs561343820 Chr.6: 35475725-35475725 on Build GRCh38
rs912648993 Chr.6: 35475767-35475767 on Build GRCh38
rs931829876 Chr.6: 35475760-35475760 on Build GRCh38
rs370153314 Chr.6: 35475730-35475730 on Build GRCh38
rs897155325 Chr.6: 35475805-35475805 on Build GRCh38
rs777123743 Chr.6: 35475706-35475706 on Build GRCh38
rs981372951 Chr.6: 35475812-35475812 on Build GRCh38
rs992013336 Chr.6: 35475671-35475671 on Build GRCh38
rs747005588 Chr.6: 35475713-35475713 on Build GRCh38
rs2076170 Chr.6: 35475796-35475796 on Build GRCh38
rs754157370 Chr.6: 35475675-35475675 on Build GRCh38
rs906369131 Chr.6: 35475787-35475787 on Build GRCh38
rs778472538 Chr.6: 35475692-35475692 on Build GRCh38
rs750488566 Chr.6: 35475708-35475709 on Build GRCh38
rs766800640 Chr.6: 35475674-35475674 on Build GRCh38
rs12524879 Chr.6: 35475720-35475720 on Build GRCh38
rs190062381 Chr.6: 35475778-35475778 on Build GRCh38
rs776590577 Chr.6: 35475721-35475721 on Build GRCh38
rs543797739 Chr.6: 35475853-35475853 on Build GRCh38
rs538245778 Chr.6: 35475752-35475752 on Build GRCh38
rs769001267 Chr.6: 35475738-35475738 on Build GRCh38
rs762110717 Chr.6: 35475756-35475756 on Build GRCh38
rs549970732 Chr.6: 35475764-35475764 on Build GRCh38
rs971674181 Chr.6: 35475802-35475802 on Build GRCh38
rs373604865 Chr.6: 35475703-35475703 on Build GRCh38
rs374944915 Chr.6: 35475704-35475704 on Build GRCh38
rs891782017 Chr.6: 35475769-35475769 on Build GRCh38

View Full Product Details