Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22885179-22885446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REM2
Entrez Gene ID
161253
Gene Name
RRAD and GEM like GTPase 2
Gene Aliases
-
UniGene
Hs.444911
Gene Chromosome Location
Chr.14: 22883165-22887680 on Build GRCh38
Transcripts
NM_173527.2 XM_005267383.3 XM_011536518.2 XM_017021058.1 XM_017021059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1369100 c.459G>T p.V153V Chr.14: 22885309-22885309 on Build GRCh38
4969474 c.434G>A p.R145H Chr.14: 22885284-22885284 on Build GRCh38
954547 c.433C>T p.R145C Chr.14: 22885283-22885283 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765682281 Chr.14: 22885273-22885273 on Build GRCh38
rs916401477 Chr.14: 22885353-22885353 on Build GRCh38
rs371952944 Chr.14: 22885247-22885247 on Build GRCh38
rs774885561 Chr.14: 22885344-22885344 on Build GRCh38
rs150780091 Chr.14: 22885373-22885373 on Build GRCh38
rs768206017 Chr.14: 22885354-22885354 on Build GRCh38
rs770774317 Chr.14: 22885301-22885301 on Build GRCh38
rs769826661 Chr.14: 22885283-22885283 on Build GRCh38
rs774218813 Chr.14: 22885235-22885235 on Build GRCh38
rs376841437 Chr.14: 22885254-22885254 on Build GRCh38
rs534333966 Chr.14: 22885256-22885256 on Build GRCh38
rs750436070 Chr.14: 22885379-22885380 on Build GRCh38
rs766865917 Chr.14: 22885379-22885379 on Build GRCh38
rs772901860 Chr.14: 22885219-22885219 on Build GRCh38
rs1052078666 Chr.14: 22885413-22885413 on Build GRCh38
rs775036316 Chr.14: 22885250-22885250 on Build GRCh38
rs781412286 Chr.14: 22885290-22885290 on Build GRCh38
rs759459921 Chr.14: 22885246-22885246 on Build GRCh38
rs939579541 Chr.14: 22885275-22885275 on Build GRCh38
rs73588590 Chr.14: 22885233-22885233 on Build GRCh38
rs557705531 Chr.14: 22885318-22885318 on Build GRCh38
rs760163746 Chr.14: 22885352-22885352 on Build GRCh38
rs755189659 Chr.14: 22885284-22885284 on Build GRCh38
rs774128911 Chr.14: 22885303-22885303 on Build GRCh38
rs749097349 Chr.14: 22885296-22885296 on Build GRCh38
rs776481994 Chr.14: 22885355-22885355 on Build GRCh38
rs200093601 Chr.14: 22885356-22885356 on Build GRCh38
rs1056719322 Chr.14: 22885277-22885277 on Build GRCh38
rs996668778 Chr.14: 22885211-22885211 on Build GRCh38
rs772024367 Chr.14: 22885327-22885327 on Build GRCh38
rs765034461 Chr.14: 22885367-22885367 on Build GRCh38
rs979226372 Chr.14: 22885365-22885365 on Build GRCh38
rs754156511 Chr.14: 22885259-22885259 on Build GRCh38
rs751037447 Chr.14: 22885276-22885276 on Build GRCh38
rs757637804 Chr.14: 22885268-22885268 on Build GRCh38
rs375658016 Chr.14: 22885217-22885217 on Build GRCh38
rs551138388 Chr.14: 22885215-22885215 on Build GRCh38
rs745969204 Chr.14: 22885205-22885205 on Build GRCh38
rs569437394 Chr.14: 22885364-22885364 on Build GRCh38
rs141462741 Chr.14: 22885372-22885372 on Build GRCh38
rs758380439 Chr.14: 22885381-22885382 on Build GRCh38
rs758940214 Chr.14: 22885279-22885279 on Build GRCh38
rs749047109 Chr.14: 22885225-22885225 on Build GRCh38
rs935064688 Chr.14: 22885401-22885401 on Build GRCh38

View Full Product Details