Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136016464-136016705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9A6
Entrez Gene ID
10479
Gene Name
solute carrier family 9 member A6
Gene Aliases
MRSA, NHE6
UniGene
Hs.62185
Gene Chromosome Location
Chr.X: 135974597-136047269 on Build GRCh38
Transcripts
NM_001042537.1 NM_001177651.1 NM_006359.2 XM_006724726.3 XM_011531243.2 XM_017029223.1 XM_017029224.1 XM_017029225.1

SNP Detail

SNP ID Chromosome Location
rs201179989 Chr.X: 136016617-136016617 on Build GRCh38
rs140158476 Chr.X: 136016683-136016683 on Build GRCh38
rs781927478 Chr.X: 136016632-136016632 on Build GRCh38
rs782367882 Chr.X: 136016630-136016630 on Build GRCh38
rs376479501 Chr.X: 136016627-136016627 on Build GRCh38
rs782204082 Chr.X: 136016645-136016645 on Build GRCh38
rs940364758 Chr.X: 136016529-136016529 on Build GRCh38
rs920490848 Chr.X: 136016587-136016587 on Build GRCh38
rs151178361 Chr.X: 136016647-136016647 on Build GRCh38
rs370038128 Chr.X: 136016583-136016583 on Build GRCh38
rs781844585 Chr.X: 136016565-136016565 on Build GRCh38

View Full Product Details