Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 42347261-42347522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFF2
Entrez Gene ID
7032
Gene Name
trefoil factor 2
Gene Aliases
SML1, SP
UniGene
Hs.2979
Gene Chromosome Location
Chr.21: 42346357-42351099 on Build GRCh38
Transcripts
NM_005423.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6800551 c.362C>T p.P121L Chr.21: 42347500-42347500 on Build GRCh38
6344695 c.346C>A p.P116T Chr.21: 42347516-42347516 on Build GRCh38
1414260 c.363G>A p.P121P Chr.21: 42347499-42347499 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs908395893 Chr.21: 42347280-42347280 on Build GRCh38
rs543277873 Chr.21: 42347450-42347450 on Build GRCh38
rs200812929 Chr.21: 42347420-42347420 on Build GRCh38
rs753779328 Chr.21: 42347441-42347441 on Build GRCh38
rs1009674050 Chr.21: 42347458-42347461 on Build GRCh38
rs758538116 Chr.21: 42347452-42347452 on Build GRCh38
rs746320531 Chr.21: 42347478-42347478 on Build GRCh38
rs1054319525 Chr.21: 42347401-42347401 on Build GRCh38
rs766307182 Chr.21: 42347440-42347440 on Build GRCh38
rs225337 Chr.21: 42347391-42347391 on Build GRCh38
rs751893244 Chr.21: 42347460-42347460 on Build GRCh38
rs182546610 Chr.21: 42347313-42347313 on Build GRCh38
rs746210135 Chr.21: 42347450-42347450 on Build GRCh38
rs574366275 Chr.21: 42347426-42347426 on Build GRCh38
rs140013939 Chr.21: 42347380-42347380 on Build GRCh38
rs765164729 Chr.21: 42347445-42347445 on Build GRCh38
rs374026310 Chr.21: 42347480-42347480 on Build GRCh38
rs998908913 Chr.21: 42347394-42347394 on Build GRCh38
rs547675942 Chr.21: 42347297-42347297 on Build GRCh38
rs73225467 Chr.21: 42347384-42347384 on Build GRCh38
rs145769308 Chr.21: 42347486-42347486 on Build GRCh38
rs1023249031 Chr.21: 42347462-42347462 on Build GRCh38
rs554464449 Chr.21: 42347398-42347398 on Build GRCh38
rs556785400 Chr.21: 42347477-42347477 on Build GRCh38
rs529863144 Chr.21: 42347309-42347309 on Build GRCh38
rs368840932 Chr.21: 42347472-42347472 on Build GRCh38
rs764349968 Chr.21: 42347458-42347458 on Build GRCh38
rs267606140 Chr.21: 42347502-42347502 on Build GRCh38
rs77037411 Chr.21: 42347314-42347314 on Build GRCh38
rs770167723 Chr.21: 42347463-42347464 on Build GRCh38
rs112275939 Chr.21: 42347310-42347310 on Build GRCh38
rs225336 Chr.21: 42347305-42347305 on Build GRCh38
rs143520404 Chr.21: 42347493-42347493 on Build GRCh38
rs757580151 Chr.21: 42347474-42347474 on Build GRCh38
rs759530532 Chr.21: 42347444-42347444 on Build GRCh38
rs538954796 Chr.21: 42347481-42347481 on Build GRCh38
rs982586399 Chr.21: 42347279-42347279 on Build GRCh38
rs896009188 Chr.21: 42347418-42347418 on Build GRCh38

View Full Product Details