Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11678989-11679250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNN
Entrez Gene ID
8303
Gene Name
stannin
Gene Aliases
-
UniGene
Hs.618526
Gene Chromosome Location
Chr.16: 11668433-11679159 on Build GRCh38
Transcripts
NM_003498.5 XM_017023741.1 XM_017023742.1 NM_001303447.1 NM_001324022.1 NM_001324024.1 NM_001324025.1 NM_015914.6 NR_136671.1 NR_136672.1 NR_136673.1 NR_136674.1 XM_011522515.2 XM_011522516.2 XM_017023268.1

Gene Symbol
TXNDC11
Entrez Gene ID
51061
Gene Name
thioredoxin domain containing 11
Gene Aliases
EFP1
UniGene
Hs.313847
Gene Chromosome Location
Chr.16: 11679080-11742878 on Build GRCh38
Transcripts
NM_003498.5 XM_017023741.1 XM_017023742.1 NM_001303447.1 NM_001324022.1 NM_001324024.1 NM_001324025.1 NM_015914.6 NR_136671.1 NR_136672.1 NR_136673.1 NR_136674.1 XM_011522515.2 XM_011522516.2 XM_017023268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3794571 c.2929G>C p.E977Q Chr.16: 11679224-11679224 on Build GRCh38
1640390 c.2906A>T p.N969I Chr.16: 11679247-11679247 on Build GRCh38
286214 c.2862G>T p.E954D Chr.16: 11679210-11679210 on Build GRCh38
4873094 c.2943G>T p.E981D Chr.16: 11679210-11679210 on Build GRCh38
5887451 c.2864A>C p.N955T Chr.16: 11679208-11679208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746114850 Chr.16: 11679181-11679181 on Build GRCh38
rs562437497 Chr.16: 11679126-11679126 on Build GRCh38
rs959958782 Chr.16: 11679079-11679079 on Build GRCh38
rs36012200 Chr.16: 11679147-11679148 on Build GRCh38
rs972161442 Chr.16: 11679050-11679050 on Build GRCh38
rs948235118 Chr.16: 11679114-11679114 on Build GRCh38
rs979561855 Chr.16: 11679138-11679138 on Build GRCh38
rs139507945 Chr.16: 11679233-11679233 on Build GRCh38
rs960523764 Chr.16: 11679063-11679063 on Build GRCh38
rs745947611 Chr.16: 11679075-11679075 on Build GRCh38
rs368797018 Chr.16: 11679203-11679203 on Build GRCh38
rs961603310 Chr.16: 11679106-11679109 on Build GRCh38
rs550070163 Chr.16: 11679047-11679047 on Build GRCh38
rs545731522 Chr.16: 11679125-11679125 on Build GRCh38
rs141628246 Chr.16: 11679191-11679191 on Build GRCh38
rs762745074 Chr.16: 11679218-11679218 on Build GRCh38
rs986177858 Chr.16: 11679171-11679171 on Build GRCh38
rs769460739 Chr.16: 11679215-11679215 on Build GRCh38
rs8191348 Chr.16: 11679097-11679097 on Build GRCh38
rs779075493 Chr.16: 11679180-11679180 on Build GRCh38
rs772120791 Chr.16: 11679108-11679108 on Build GRCh38
rs752000865 Chr.16: 11679182-11679184 on Build GRCh38
rs755526872 Chr.16: 11679193-11679196 on Build GRCh38
rs773264602 Chr.16: 11679216-11679216 on Build GRCh38
rs1013446119 Chr.16: 11679096-11679096 on Build GRCh38
rs920592963 Chr.16: 11679130-11679130 on Build GRCh38
rs780104836 Chr.16: 11679200-11679200 on Build GRCh38
rs952114636 Chr.16: 11679170-11679170 on Build GRCh38
rs1015506281 Chr.16: 11679105-11679105 on Build GRCh38
rs757416382 Chr.16: 11679162-11679162 on Build GRCh38
rs767708463 Chr.16: 11679210-11679212 on Build GRCh38
rs370523801 Chr.16: 11679027-11679027 on Build GRCh38

View Full Product Details