Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132271734-132271991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDLIM4
Entrez Gene ID
8572
Gene Name
PDZ and LIM domain 4
Gene Aliases
RIL
UniGene
Hs.424312
Gene Chromosome Location
Chr.5: 132257658-132273454 on Build GRCh38
Transcripts
NM_001131027.1 NM_003687.3 XM_017010002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3393121 c.713C>T p.T238M Chr.5: 132271833-132271833 on Build GRCh38
5780249 c.776G>T p.G259V Chr.5: 132271896-132271896 on Build GRCh38
3137245 c.730G>A p.A244T Chr.5: 132271850-132271850 on Build GRCh38
6743371 c.774C>T p.C258C Chr.5: 132271894-132271894 on Build GRCh38
7240160 c.690C>A p.G230G Chr.5: 132271810-132271810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768047246 Chr.5: 132271849-132271849 on Build GRCh38
rs371120846 Chr.5: 132271834-132271834 on Build GRCh38
rs755069170 Chr.5: 132271869-132271869 on Build GRCh38
rs1056815475 Chr.5: 132271940-132271940 on Build GRCh38
rs1002864280 Chr.5: 132271966-132271966 on Build GRCh38
rs868361592 Chr.5: 132271791-132271791 on Build GRCh38
rs780844524 Chr.5: 132271773-132271773 on Build GRCh38
rs775352397 Chr.5: 132271847-132271847 on Build GRCh38
rs776588349 Chr.5: 132271835-132271835 on Build GRCh38
rs201790154 Chr.5: 132271942-132271942 on Build GRCh38
rs990621447 Chr.5: 132271758-132271758 on Build GRCh38
rs748813165 Chr.5: 132271959-132271959 on Build GRCh38
rs781012152 Chr.5: 132271941-132271941 on Build GRCh38
rs1042146360 Chr.5: 132271802-132271802 on Build GRCh38
rs557597794 Chr.5: 132271824-132271824 on Build GRCh38
rs535617458 Chr.5: 132271761-132271761 on Build GRCh38
rs765348419 Chr.5: 132271921-132271921 on Build GRCh38
rs760005543 Chr.5: 132271919-132271919 on Build GRCh38
rs201243327 Chr.5: 132271814-132271814 on Build GRCh38
rs162884 Chr.5: 132271801-132271801 on Build GRCh38
rs758970379 Chr.5: 132271840-132271840 on Build GRCh38
rs4877 Chr.5: 132271895-132271895 on Build GRCh38
rs942811804 Chr.5: 132271854-132271854 on Build GRCh38
rs563176113 Chr.5: 132271970-132271970 on Build GRCh38
rs767992186 Chr.5: 132271786-132271786 on Build GRCh38
rs879454942 Chr.5: 132271793-132271793 on Build GRCh38
rs758428933 Chr.5: 132271928-132271928 on Build GRCh38
rs200399687 Chr.5: 132271772-132271772 on Build GRCh38
rs780562316 Chr.5: 132271808-132271808 on Build GRCh38
rs747013369 Chr.5: 132271828-132271828 on Build GRCh38
rs752327613 Chr.5: 132271777-132271777 on Build GRCh38
rs750840954 Chr.5: 132271855-132271855 on Build GRCh38
rs771245370 Chr.5: 132271858-132271879 on Build GRCh38
rs777973438 Chr.5: 132271935-132271935 on Build GRCh38
rs866990886 Chr.5: 132271788-132271788 on Build GRCh38
rs752956503 Chr.5: 132271879-132271879 on Build GRCh38
rs555841710 Chr.5: 132271762-132271762 on Build GRCh38
rs757758144 Chr.5: 132271781-132271781 on Build GRCh38
rs758595611 Chr.5: 132271886-132271886 on Build GRCh38
rs781699893 Chr.5: 132271873-132271915 on Build GRCh38
rs766802234 Chr.5: 132271862-132271862 on Build GRCh38
rs751011653 Chr.5: 132271799-132271799 on Build GRCh38
rs780099459 Chr.5: 132271956-132271956 on Build GRCh38
rs371367801 Chr.5: 132271901-132271901 on Build GRCh38
rs762507226 Chr.5: 132271900-132271900 on Build GRCh38
rs766751359 Chr.5: 132271907-132271907 on Build GRCh38
rs760962631 Chr.5: 132271904-132271904 on Build GRCh38
rs751744427 Chr.5: 132271936-132271936 on Build GRCh38
rs374657990 Chr.5: 132271903-132271903 on Build GRCh38
rs769879591 Chr.5: 132271952-132271952 on Build GRCh38
rs754367110 Chr.5: 132271918-132271918 on Build GRCh38
rs374068089 Chr.5: 132271887-132271887 on Build GRCh38
rs771921138 Chr.5: 132271822-132271822 on Build GRCh38
rs748354504 Chr.5: 132271819-132271819 on Build GRCh38
rs770691527 Chr.5: 132271890-132271890 on Build GRCh38
rs368641903 Chr.5: 132271938-132271938 on Build GRCh38
rs778957379 Chr.5: 132271877-132271877 on Build GRCh38
rs375520975 Chr.5: 132271818-132271818 on Build GRCh38
rs752792890 Chr.5: 132271922-132271922 on Build GRCh38
rs775122958 Chr.5: 132271897-132271897 on Build GRCh38
rs781003083 Chr.5: 132271894-132271894 on Build GRCh38
rs746807572 Chr.5: 132271889-132271889 on Build GRCh38
rs755020798 Chr.5: 132271816-132271816 on Build GRCh38
rs769418342 Chr.5: 132271843-132271843 on Build GRCh38

View Full Product Details