Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129719074-129719307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRX2
Entrez Gene ID
51450
Gene Name
paired related homeobox 2
Gene Aliases
PMX2, PRX2
UniGene
Hs.660115
Gene Chromosome Location
Chr.9: 129665641-129722674 on Build GRCh38
Transcripts
NM_016307.3 XM_017014803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4138856 c.260-9C>A p.? Chr.9: 129719222-129719222 on Build GRCh38
1314555 c.278G>A p.G93E Chr.9: 129719249-129719249 on Build GRCh38
6757124 c.322C>T p.R108C Chr.9: 129719293-129719293 on Build GRCh38
6425067 c.271A>G p.S91G Chr.9: 129719242-129719242 on Build GRCh38
3328469 c.289G>A p.A97T Chr.9: 129719260-129719260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527354835 Chr.9: 129719263-129719263 on Build GRCh38
rs201665194 Chr.9: 129719274-129719274 on Build GRCh38
rs200713340 Chr.9: 129719217-129719218 on Build GRCh38
rs573508810 Chr.9: 129719131-129719131 on Build GRCh38
rs149764500 Chr.9: 129719283-129719283 on Build GRCh38
rs771071390 Chr.9: 129719217-129719217 on Build GRCh38
rs979568870 Chr.9: 129719128-129719128 on Build GRCh38
rs564207520 Chr.9: 129719214-129719215 on Build GRCh38
rs553501537 Chr.9: 129719122-129719122 on Build GRCh38
rs772088334 Chr.9: 129719214-129719214 on Build GRCh38
rs754586483 Chr.9: 129719209-129719209 on Build GRCh38
rs770826522 Chr.9: 129719270-129719270 on Build GRCh38
rs371032092 Chr.9: 129719218-129719218 on Build GRCh38
rs141641901 Chr.9: 129719164-129719164 on Build GRCh38
rs935383203 Chr.9: 129719129-129719129 on Build GRCh38
rs370204978 Chr.9: 129719265-129719265 on Build GRCh38
rs575925541 Chr.9: 129719185-129719185 on Build GRCh38
rs370963746 Chr.9: 129719137-129719137 on Build GRCh38
rs3214712 Chr.9: 129719212-129719213 on Build GRCh38
rs775573934 Chr.9: 129719288-129719288 on Build GRCh38
rs925350899 Chr.9: 129719128-129719128 on Build GRCh38
rs377627645 Chr.9: 129719195-129719195 on Build GRCh38
rs530396570 Chr.9: 129719215-129719215 on Build GRCh38
rs754567369 Chr.9: 129719247-129719247 on Build GRCh38
rs777230786 Chr.9: 129719246-129719246 on Build GRCh38
rs763470813 Chr.9: 129719219-129719219 on Build GRCh38
rs1049559282 Chr.9: 129719168-129719168 on Build GRCh38
rs564891395 Chr.9: 129719235-129719235 on Build GRCh38
rs748147016 Chr.9: 129719211-129719211 on Build GRCh38
rs969526097 Chr.9: 129719116-129719116 on Build GRCh38
rs767772157 Chr.9: 129719243-129719243 on Build GRCh38
rs896914944 Chr.9: 129719221-129719221 on Build GRCh38
rs751921861 Chr.9: 129719234-129719234 on Build GRCh38
rs755749704 Chr.9: 129719182-129719182 on Build GRCh38
rs35865713 Chr.9: 129719268-129719268 on Build GRCh38
rs757564666 Chr.9: 129719218-129719219 on Build GRCh38
rs994213997 Chr.9: 129719239-129719239 on Build GRCh38
rs778258722 Chr.9: 129719210-129719210 on Build GRCh38
rs765552639 Chr.9: 129719213-129719213 on Build GRCh38
rs1026609785 Chr.9: 129719277-129719277 on Build GRCh38
rs369683267 Chr.9: 129719216-129719217 on Build GRCh38
rs397948759 Chr.9: 129719219-129719220 on Build GRCh38
rs752216948 Chr.9: 129719264-129719264 on Build GRCh38
rs746928887 Chr.9: 129719269-129719269 on Build GRCh38
rs201179051 Chr.9: 129719278-129719278 on Build GRCh38
rs769856483 Chr.9: 129719287-129719287 on Build GRCh38
rs779702780 Chr.9: 129719192-129719192 on Build GRCh38
rs188364858 Chr.9: 129719114-129719114 on Build GRCh38
rs913156807 Chr.9: 129719130-129719130 on Build GRCh38

View Full Product Details