Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74429605-74429870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTKN
Entrez Gene ID
6242
Gene Name
rhotekin
Gene Aliases
-
UniGene
Hs.192854
Gene Chromosome Location
Chr.2: 74425861-74441933 on Build GRCh38
Transcripts
NM_001015055.1 NM_001015056.1 NM_033046.2 XM_005264478.2 XM_005264479.4 XM_017004635.1 XM_017004636.1 XM_017004637.1 XM_017004638.1 XM_017004639.1 XM_017004640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5397420 c.680G>A p.G227E Chr.2: 74429864-74429864 on Build GRCh38
6770330 c.686G>A p.S229N Chr.2: 74429858-74429858 on Build GRCh38
257926 c.722G>A p.S241N Chr.2: 74429861-74429861 on Build GRCh38
3372871 c.752T>C p.V251A Chr.2: 74429831-74429831 on Build GRCh38
4949866 c.727C>T p.P243S Chr.2: 74429856-74429856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037147490 Chr.2: 74429633-74429633 on Build GRCh38
rs780574127 Chr.2: 74429848-74429848 on Build GRCh38
rs763095611 Chr.2: 74429664-74429664 on Build GRCh38
rs980491340 Chr.2: 74429750-74429750 on Build GRCh38
rs1006940886 Chr.2: 74429672-74429672 on Build GRCh38
rs774605770 Chr.2: 74429805-74429805 on Build GRCh38
rs910162183 Chr.2: 74429764-74429764 on Build GRCh38
rs750459053 Chr.2: 74429823-74429823 on Build GRCh38
rs762053320 Chr.2: 74429821-74429821 on Build GRCh38
rs1040913284 Chr.2: 74429753-74429753 on Build GRCh38
rs1043669310 Chr.2: 74429676-74429677 on Build GRCh38
rs917240 Chr.2: 74429729-74429729 on Build GRCh38
rs770272522 Chr.2: 74429796-74429796 on Build GRCh38
rs574306612 Chr.2: 74429806-74429806 on Build GRCh38
rs534474599 Chr.2: 74429743-74429743 on Build GRCh38
rs566747314 Chr.2: 74429733-74429733 on Build GRCh38
rs777259471 Chr.2: 74429789-74429789 on Build GRCh38
rs745347530 Chr.2: 74429850-74429850 on Build GRCh38
rs972913600 Chr.2: 74429807-74429807 on Build GRCh38
rs139676001 Chr.2: 74429845-74429845 on Build GRCh38
rs569000574 Chr.2: 74429689-74429689 on Build GRCh38
rs746657691 Chr.2: 74429840-74429840 on Build GRCh38
rs777439644 Chr.2: 74429839-74429839 on Build GRCh38
rs78167177 Chr.2: 74429702-74429702 on Build GRCh38
rs934121353 Chr.2: 74429836-74429836 on Build GRCh38
rs777255641 Chr.2: 74429680-74429680 on Build GRCh38
rs991154641 Chr.2: 74429690-74429690 on Build GRCh38
rs778870230 Chr.2: 74429832-74429832 on Build GRCh38
rs915659367 Chr.2: 74429697-74429697 on Build GRCh38
rs754873936 Chr.2: 74429831-74429831 on Build GRCh38
rs933750891 Chr.2: 74429649-74429649 on Build GRCh38
rs374352210 Chr.2: 74429802-74429802 on Build GRCh38
rs999390118 Chr.2: 74429687-74429687 on Build GRCh38
rs368296022 Chr.2: 74429822-74429822 on Build GRCh38
rs752452882 Chr.2: 74429834-74429834 on Build GRCh38
rs751800742 Chr.2: 74429814-74429814 on Build GRCh38
rs559615441 Chr.2: 74429708-74429708 on Build GRCh38
rs529429292 Chr.2: 74429643-74429643 on Build GRCh38
rs550607980 Chr.2: 74429683-74429683 on Build GRCh38
rs932615750 Chr.2: 74429795-74429795 on Build GRCh38
rs1035283719 Chr.2: 74429688-74429688 on Build GRCh38
rs758162650 Chr.2: 74429838-74429838 on Build GRCh38
rs555772087 Chr.2: 74429793-74429793 on Build GRCh38
rs759976848 Chr.2: 74429791-74429791 on Build GRCh38
rs1051244572 Chr.2: 74429656-74429656 on Build GRCh38
rs918368234 Chr.2: 74429636-74429636 on Build GRCh38

View Full Product Details