Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28328613-28328846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT20
Entrez Gene ID
90410
Gene Name
intraflagellar transport 20
Gene Aliases
-
UniGene
Hs.705431
Gene Chromosome Location
Chr.17: 28328325-28335489 on Build GRCh38
Transcripts
NM_001267774.1 NM_001267775.1 NM_001267776.1 NM_001267777.1 NM_001267778.1 NM_174887.3 NM_014573.2 XM_005257965.3

Gene Symbol
TMEM97
Entrez Gene ID
27346
Gene Name
transmembrane protein 97
Gene Aliases
MAC30
UniGene
Hs.199695
Gene Chromosome Location
Chr.17: 28319041-28328685 on Build GRCh38
Transcripts
NM_001267774.1 NM_001267775.1 NM_001267776.1 NM_001267777.1 NM_001267778.1 NM_174887.3 NM_014573.2 XM_005257965.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1646776 c.412G>A p.E138K Chr.17: 28328717-28328717 on Build GRCh38
705860 c.447A>T p.*149C Chr.17: 28328722-28328722 on Build GRCh38
4860566 c.407A>T p.E136V Chr.17: 28328722-28328722 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781983819 Chr.17: 28328698-28328698 on Build GRCh38
rs372746932 Chr.17: 28328728-28328728 on Build GRCh38
rs782751120 Chr.17: 28328755-28328755 on Build GRCh38
rs782127891 Chr.17: 28328806-28328806 on Build GRCh38
rs547109598 Chr.17: 28328757-28328757 on Build GRCh38
rs11871371 Chr.17: 28328682-28328682 on Build GRCh38
rs944080087 Chr.17: 28328684-28328684 on Build GRCh38
rs782667917 Chr.17: 28328709-28328709 on Build GRCh38
rs782338973 Chr.17: 28328705-28328705 on Build GRCh38
rs782452966 Chr.17: 28328766-28328766 on Build GRCh38
rs782049383 Chr.17: 28328726-28328726 on Build GRCh38
rs200794627 Chr.17: 28328721-28328721 on Build GRCh38
rs782096680 Chr.17: 28328700-28328700 on Build GRCh38
rs1040247510 Chr.17: 28328720-28328720 on Build GRCh38
rs782273317 Chr.17: 28328678-28328678 on Build GRCh38
rs1050227664 Chr.17: 28328749-28328749 on Build GRCh38
rs782312110 Chr.17: 28328640-28328640 on Build GRCh38
rs781876579 Chr.17: 28328743-28328743 on Build GRCh38
rs376265046 Chr.17: 28328752-28328752 on Build GRCh38
rs782041220 Chr.17: 28328643-28328643 on Build GRCh38
rs781831313 Chr.17: 28328759-28328759 on Build GRCh38
rs952917251 Chr.17: 28328670-28328670 on Build GRCh38
rs782585642 Chr.17: 28328785-28328785 on Build GRCh38
rs781809757 Chr.17: 28328736-28328737 on Build GRCh38
rs782443112 Chr.17: 28328761-28328762 on Build GRCh38
rs982925150 Chr.17: 28328675-28328675 on Build GRCh38

View Full Product Details