Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231807262-231807523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS7B
Entrez Gene ID
64708
Gene Name
COP9 signalosome subunit 7B
Gene Aliases
CSN7B, SGN7b
UniGene
Hs.335061
Gene Chromosome Location
Chr.2: 231781671-231809253 on Build GRCh38
Transcripts
NM_001282949.2 NM_001282950.2 NM_001282951.2 NM_001282952.2 NM_001308381.1 NM_022730.3 XM_006712693.1 XM_011511638.1 XM_011511639.1 XM_011511640.1 XM_017004716.1 XM_017004717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
210375 c.670G>A p.A224T Chr.2: 231807520-231807520 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748584348 Chr.2: 231807457-231807457 on Build GRCh38
rs779814575 Chr.2: 231807483-231807483 on Build GRCh38
rs770366244 Chr.2: 231807471-231807471 on Build GRCh38
rs147225626 Chr.2: 231807476-231807476 on Build GRCh38
rs983101034 Chr.2: 231807387-231807387 on Build GRCh38
rs749827014 Chr.2: 231807448-231807448 on Build GRCh38
rs752615287 Chr.2: 231807480-231807480 on Build GRCh38
rs545491432 Chr.2: 231807307-231807307 on Build GRCh38
rs79703710 Chr.2: 231807319-231807319 on Build GRCh38
rs200087852 Chr.2: 231807489-231807489 on Build GRCh38
rs191937623 Chr.2: 231807454-231807454 on Build GRCh38
rs769220277 Chr.2: 231807451-231807451 on Build GRCh38
rs563501726 Chr.2: 231807314-231807314 on Build GRCh38
rs772463695 Chr.2: 231807459-231807459 on Build GRCh38
rs991245967 Chr.2: 231807325-231807325 on Build GRCh38
rs747281710 Chr.2: 231807317-231807317 on Build GRCh38
rs943958166 Chr.2: 231807397-231807397 on Build GRCh38
rs374122300 Chr.2: 231807447-231807447 on Build GRCh38
rs187402170 Chr.2: 231807388-231807388 on Build GRCh38
rs1039364899 Chr.2: 231807431-231807431 on Build GRCh38
rs370616518 Chr.2: 231807472-231807472 on Build GRCh38
rs929847433 Chr.2: 231807384-231807384 on Build GRCh38
rs757434810 Chr.2: 231807423-231807426 on Build GRCh38
rs998664495 Chr.2: 231807315-231807315 on Build GRCh38
rs765223778 Chr.2: 231807479-231807479 on Build GRCh38
rs903030531 Chr.2: 231807313-231807313 on Build GRCh38
rs760339268 Chr.2: 231807469-231807469 on Build GRCh38
rs772783538 Chr.2: 231807461-231807461 on Build GRCh38
rs1021517451 Chr.2: 231807294-231807294 on Build GRCh38

View Full Product Details