Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74698250-74698475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300LF
Entrez Gene ID
146722
Gene Name
CD300 molecule like family member f
Gene Aliases
CD300f, CLM-1, CLM1, IREM-1, IREM1, IgSF13, LMIR3, NKIR
UniGene
Hs.567706
Gene Chromosome Location
Chr.17: 74694308-74713059 on Build GRCh38
Transcripts
NM_001289082.1 NM_001289083.1 NM_001289084.1 NM_001289085.1 NM_001289086.1 NM_001289087.1 NM_139018.4 NR_110298.1 XM_017024212.1 XM_017024213.1 XM_017024214.1 XM_017024215.1 XM_017024216.1 XM_017024217.1 NM_175738.4

Gene Symbol
RAB37
Entrez Gene ID
326624
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.351413
Gene Chromosome Location
Chr.17: 74671117-74747335 on Build GRCh38
Transcripts
NM_001289082.1 NM_001289083.1 NM_001289084.1 NM_001289085.1 NM_001289086.1 NM_001289087.1 NM_139018.4 NR_110298.1 XM_017024212.1 XM_017024213.1 XM_017024214.1 XM_017024215.1 XM_017024216.1 XM_017024217.1 NM_175738.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7109612 c.506T>G p.L169R Chr.17: 74698422-74698422 on Build GRCh38
6433566 c.544A>C p.K182Q Chr.17: 74698384-74698384 on Build GRCh38
7109614 c.560T>G p.L187R Chr.17: 74698422-74698422 on Build GRCh38
6147994 c.551T>A p.L184* Chr.17: 74698431-74698431 on Build GRCh38
4786084 c.570C>T p.A190A Chr.17: 74698412-74698412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371420753 Chr.17: 74698357-74698357 on Build GRCh38
rs897956988 Chr.17: 74698301-74698301 on Build GRCh38
rs1049756137 Chr.17: 74698337-74698337 on Build GRCh38
rs867433005 Chr.17: 74698436-74698436 on Build GRCh38
rs1050521840 Chr.17: 74698270-74698270 on Build GRCh38
rs144781345 Chr.17: 74698377-74698377 on Build GRCh38
rs944887555 Chr.17: 74698296-74698296 on Build GRCh38
rs762046788 Chr.17: 74698448-74698448 on Build GRCh38
rs761033751 Chr.17: 74698344-74698344 on Build GRCh38
rs745858615 Chr.17: 74698405-74698405 on Build GRCh38
rs776704853 Chr.17: 74698330-74698330 on Build GRCh38
rs530153186 Chr.17: 74698361-74698361 on Build GRCh38
rs1025255100 Chr.17: 74698302-74698302 on Build GRCh38
rs775639502 Chr.17: 74698331-74698331 on Build GRCh38
rs777096299 Chr.17: 74698445-74698445 on Build GRCh38
rs148594652 Chr.17: 74698411-74698411 on Build GRCh38
rs749983019 Chr.17: 74698359-74698359 on Build GRCh38
rs1032015009 Chr.17: 74698380-74698380 on Build GRCh38
rs750842364 Chr.17: 74698453-74698453 on Build GRCh38
rs75145504 Chr.17: 74698294-74698294 on Build GRCh38
rs375470682 Chr.17: 74698348-74698348 on Build GRCh38
rs759502210 Chr.17: 74698319-74698319 on Build GRCh38
rs113495072 Chr.17: 74698338-74698338 on Build GRCh38
rs532653695 Chr.17: 74698450-74698450 on Build GRCh38
rs191687260 Chr.17: 74698303-74698303 on Build GRCh38
rs746589516 Chr.17: 74698385-74698385 on Build GRCh38
rs199831669 Chr.17: 74698386-74698386 on Build GRCh38
rs201214384 Chr.17: 74698432-74698432 on Build GRCh38
rs757833040 Chr.17: 74698353-74698353 on Build GRCh38
rs1055059726 Chr.17: 74698281-74698281 on Build GRCh38
rs149661344 Chr.17: 74698435-74698435 on Build GRCh38
rs765634345 Chr.17: 74698355-74698355 on Build GRCh38
rs1040542283 Chr.17: 74698299-74698299 on Build GRCh38
rs754141944 Chr.17: 74698350-74698350 on Build GRCh38
rs111797936 Chr.17: 74698412-74698412 on Build GRCh38
rs760947857 Chr.17: 74698434-74698434 on Build GRCh38
rs200130350 Chr.17: 74698439-74698439 on Build GRCh38
rs561283233 Chr.17: 74698347-74698347 on Build GRCh38
rs35919215 Chr.17: 74698417-74698417 on Build GRCh38
rs1033424531 Chr.17: 74698392-74698392 on Build GRCh38
rs1001812803 Chr.17: 74698388-74698388 on Build GRCh38
rs985065044 Chr.17: 74698398-74698398 on Build GRCh38

View Full Product Details