Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 38396315-38396570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1B1
Entrez Gene ID
219
Gene Name
aldehyde dehydrogenase 1 family member B1
Gene Aliases
ALDH5, ALDHX
UniGene
Hs.436219
Gene Chromosome Location
Chr.9: 38392664-38398665 on Build GRCh38
Transcripts
NM_000692.4 XM_011517802.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598214 c.600C>T p.L200L Chr.9: 38396348-38396348 on Build GRCh38
254510 c.652G>T p.A218S Chr.9: 38396400-38396400 on Build GRCh38
3943039 c.748G>A p.A250T Chr.9: 38396496-38396496 on Build GRCh38
6851224 c.757G>A p.V253M Chr.9: 38396505-38396505 on Build GRCh38
5965780 c.648C>G p.L216L Chr.9: 38396396-38396396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201489278 Chr.9: 38396341-38396341 on Build GRCh38
rs762792748 Chr.9: 38396495-38396495 on Build GRCh38
rs551776658 Chr.9: 38396460-38396460 on Build GRCh38
rs779633928 Chr.9: 38396464-38396464 on Build GRCh38
rs757657775 Chr.9: 38396463-38396463 on Build GRCh38
rs777800823 Chr.9: 38396537-38396537 on Build GRCh38
rs766680571 Chr.9: 38396415-38396415 on Build GRCh38
rs557733010 Chr.9: 38396421-38396421 on Build GRCh38
rs1020361812 Chr.9: 38396434-38396434 on Build GRCh38
rs767845549 Chr.9: 38396503-38396503 on Build GRCh38
rs748412978 Chr.9: 38396474-38396474 on Build GRCh38
rs966425990 Chr.9: 38396452-38396452 on Build GRCh38
rs371750489 Chr.9: 38396348-38396348 on Build GRCh38
rs771700240 Chr.9: 38396364-38396364 on Build GRCh38
rs769834866 Chr.9: 38396488-38396488 on Build GRCh38
rs4646773 Chr.9: 38396353-38396353 on Build GRCh38
rs71504569 Chr.9: 38396391-38396391 on Build GRCh38
rs770780314 Chr.9: 38396541-38396541 on Build GRCh38
rs572997069 Chr.9: 38396444-38396444 on Build GRCh38
rs367927705 Chr.9: 38396342-38396342 on Build GRCh38
rs780948768 Chr.9: 38396529-38396529 on Build GRCh38
rs780563987 Chr.9: 38396471-38396471 on Build GRCh38
rs767506066 Chr.9: 38396435-38396435 on Build GRCh38
rs754577203 Chr.9: 38396516-38396516 on Build GRCh38
rs769922673 Chr.9: 38396401-38396401 on Build GRCh38
rs759678392 Chr.9: 38396501-38396501 on Build GRCh38
rs372171593 Chr.9: 38396453-38396453 on Build GRCh38
rs905064991 Chr.9: 38396412-38396412 on Build GRCh38
rs200322513 Chr.9: 38396394-38396394 on Build GRCh38
rs773129997 Chr.9: 38396405-38396405 on Build GRCh38
rs755719695 Chr.9: 38396532-38396532 on Build GRCh38
rs762893637 Chr.9: 38396414-38396414 on Build GRCh38
rs759608147 Chr.9: 38396425-38396425 on Build GRCh38
rs747754718 Chr.9: 38396479-38396479 on Build GRCh38
rs778113285 Chr.9: 38396357-38396357 on Build GRCh38
rs774503656 Chr.9: 38396424-38396424 on Build GRCh38
rs941162320 Chr.9: 38396389-38396389 on Build GRCh38
rs199527495 Chr.9: 38396349-38396349 on Build GRCh38
rs754365863 Chr.9: 38396509-38396509 on Build GRCh38
rs4878199 Chr.9: 38396505-38396505 on Build GRCh38
rs772132330 Chr.9: 38396525-38396526 on Build GRCh38
rs200179372 Chr.9: 38396451-38396451 on Build GRCh38
rs145426005 Chr.9: 38396375-38396375 on Build GRCh38
rs749718210 Chr.9: 38396363-38396363 on Build GRCh38
rs768074734 Chr.9: 38396392-38396392 on Build GRCh38
rs753178763 Chr.9: 38396442-38396442 on Build GRCh38
rs150776963 Chr.9: 38396531-38396531 on Build GRCh38
rs757559755 Chr.9: 38396511-38396511 on Build GRCh38
rs768147935 Chr.9: 38396469-38396469 on Build GRCh38
rs770868005 Chr.9: 38396496-38396496 on Build GRCh38
rs201458895 Chr.9: 38396512-38396512 on Build GRCh38
rs779188273 Chr.9: 38396456-38396456 on Build GRCh38
rs750868385 Chr.9: 38396514-38396514 on Build GRCh38
rs746656134 Chr.9: 38396465-38396465 on Build GRCh38
rs774572359 Chr.9: 38396499-38396499 on Build GRCh38
rs749247146 Chr.9: 38396540-38396540 on Build GRCh38
rs375754622 Chr.9: 38396489-38396489 on Build GRCh38
rs752704133 Chr.9: 38396504-38396504 on Build GRCh38
rs764552270 Chr.9: 38396506-38396506 on Build GRCh38
rs749435661 Chr.9: 38396363-38396364 on Build GRCh38
rs1002348391 Chr.9: 38396437-38396437 on Build GRCh38
rs759954510 Chr.9: 38396388-38396389 on Build GRCh38

View Full Product Details