Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113413466-113413731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
UniGene
Hs.632691
Gene Chromosome Location
Chr.9: 113410644-113429684 on Build GRCh38
Transcripts
NM_001278629.1 NM_001278630.1 NM_152786.2 XM_011518477.1 XM_011518478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3925977 c.3G>A p.M1I Chr.9: 113413496-113413496 on Build GRCh38
6876002 c.137C>T p.P46L Chr.9: 113413630-113413630 on Build GRCh38
119663 c.30C>G p.D10E Chr.9: 113413523-113413523 on Build GRCh38
6876003 c.138C>T p.P46P Chr.9: 113413631-113413631 on Build GRCh38
4163236 c.104A>T p.H35L Chr.9: 113413597-113413597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138974443 Chr.9: 113413502-113413502 on Build GRCh38
rs1050885321 Chr.9: 113413582-113413582 on Build GRCh38
rs892765376 Chr.9: 113413593-113413593 on Build GRCh38
rs367881038 Chr.9: 113413574-113413574 on Build GRCh38
rs763996934 Chr.9: 113413523-113413523 on Build GRCh38
rs772199543 Chr.9: 113413610-113413610 on Build GRCh38
rs772989087 Chr.9: 113413599-113413599 on Build GRCh38
rs35171810 Chr.9: 113413566-113413567 on Build GRCh38
rs766245523 Chr.9: 113413694-113413697 on Build GRCh38
rs778187103 Chr.9: 113413584-113413584 on Build GRCh38
rs756627091 Chr.9: 113413581-113413581 on Build GRCh38
rs762605848 Chr.9: 113413642-113413642 on Build GRCh38
rs371996976 Chr.9: 113413688-113413688 on Build GRCh38
rs1039244418 Chr.9: 113413575-113413575 on Build GRCh38
rs775153528 Chr.9: 113413506-113413506 on Build GRCh38
rs764961326 Chr.9: 113413541-113413541 on Build GRCh38
rs536455879 Chr.9: 113413670-113413670 on Build GRCh38
rs756867505 Chr.9: 113413532-113413532 on Build GRCh38
rs367568001 Chr.9: 113413602-113413602 on Build GRCh38
rs995677110 Chr.9: 113413698-113413698 on Build GRCh38
rs753395769 Chr.9: 113413530-113413530 on Build GRCh38
rs200426143 Chr.9: 113413655-113413655 on Build GRCh38
rs760728739 Chr.9: 113413668-113413669 on Build GRCh38
rs759778993 Chr.9: 113413493-113413493 on Build GRCh38
rs778907937 Chr.9: 113413679-113413679 on Build GRCh38
rs761863039 Chr.9: 113413630-113413630 on Build GRCh38
rs140393057 Chr.9: 113413578-113413578 on Build GRCh38
rs186126933 Chr.9: 113413491-113413491 on Build GRCh38
rs754306786 Chr.9: 113413657-113413657 on Build GRCh38
rs548386317 Chr.9: 113413624-113413624 on Build GRCh38
rs200045945 Chr.9: 113413654-113413654 on Build GRCh38
rs372019948 Chr.9: 113413611-113413611 on Build GRCh38
rs766001704 Chr.9: 113413653-113413653 on Build GRCh38
rs768605987 Chr.9: 113413616-113413616 on Build GRCh38
rs770952202 Chr.9: 113413599-113413599 on Build GRCh38
rs757769945 Chr.9: 113413676-113413676 on Build GRCh38
rs749954086 Chr.9: 113413636-113413636 on Build GRCh38
rs776757925 Chr.9: 113413628-113413628 on Build GRCh38
rs995450793 Chr.9: 113413697-113413697 on Build GRCh38
rs1027233555 Chr.9: 113413703-113413703 on Build GRCh38
rs375354869 Chr.9: 113413632-113413632 on Build GRCh38
rs749352310 Chr.9: 113413591-113413591 on Build GRCh38
rs768544169 Chr.9: 113413595-113413595 on Build GRCh38
rs773877768 Chr.9: 113413603-113413603 on Build GRCh38

View Full Product Details