Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130665564-130665825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENOX2
Entrez Gene ID
10495
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Gene Aliases
APK1, COVA1, tNOX
UniGene
Hs.171458
Gene Chromosome Location
Chr.X: 130622330-130903317 on Build GRCh38
Transcripts
NM_001281736.1 NM_006375.3 NM_182314.2 XM_005262354.3 XM_011531244.2 XM_011531245.2 XM_011531247.2 XM_011531249.2 XM_011531251.2 XM_017029226.1 XM_017029227.1 XM_017029228.1 XM_017029229.1

SNP Detail

SNP ID Chromosome Location
rs371814711 Chr.X: 130665607-130665607 on Build GRCh38
rs776993717 Chr.X: 130665727-130665727 on Build GRCh38
rs148674081 Chr.X: 130665791-130665791 on Build GRCh38
rs766732874 Chr.X: 130665625-130665625 on Build GRCh38
rs779614798 Chr.X: 130665595-130665595 on Build GRCh38
rs749365956 Chr.X: 130665720-130665720 on Build GRCh38
rs1012842936 Chr.X: 130665635-130665635 on Build GRCh38
rs146487391 Chr.X: 130665721-130665721 on Build GRCh38
rs779849967 Chr.X: 130665664-130665664 on Build GRCh38
rs370502329 Chr.X: 130665611-130665611 on Build GRCh38
rs760991786 Chr.X: 130665622-130665622 on Build GRCh38
rs750349566 Chr.X: 130665762-130665762 on Build GRCh38
rs201760447 Chr.X: 130665626-130665626 on Build GRCh38
rs759818745 Chr.X: 130665728-130665728 on Build GRCh38
rs765414849 Chr.X: 130665634-130665634 on Build GRCh38
rs190303089 Chr.X: 130665694-130665694 on Build GRCh38
rs182412471 Chr.X: 130665606-130665606 on Build GRCh38
rs773794040 Chr.X: 130665610-130665610 on Build GRCh38
rs751267883 Chr.X: 130665609-130665609 on Build GRCh38
rs370936433 Chr.X: 130665775-130665775 on Build GRCh38
rs775835134 Chr.X: 130665731-130665731 on Build GRCh38
rs780666351 Chr.X: 130665753-130665753 on Build GRCh38
rs760605094 Chr.X: 130665774-130665774 on Build GRCh38
rs766604848 Chr.X: 130665607-130665607 on Build GRCh38
rs377683557 Chr.X: 130665700-130665700 on Build GRCh38
rs759750848 Chr.X: 130665628-130665628 on Build GRCh38
rs747712251 Chr.X: 130665708-130665708 on Build GRCh38
rs770203721 Chr.X: 130665729-130665729 on Build GRCh38
rs755919115 Chr.X: 130665594-130665594 on Build GRCh38
rs142549504 Chr.X: 130665658-130665658 on Build GRCh38
rs763168882 Chr.X: 130665747-130665747 on Build GRCh38
rs752895702 Chr.X: 130665645-130665645 on Build GRCh38
rs187017146 Chr.X: 130665613-130665613 on Build GRCh38
rs753732518 Chr.X: 130665776-130665776 on Build GRCh38
rs755894800 Chr.X: 130665663-130665663 on Build GRCh38
rs754590913 Chr.X: 130665697-130665697 on Build GRCh38
rs760816362 Chr.X: 130665606-130665607 on Build GRCh38
rs771218552 Chr.X: 130665725-130665725 on Build GRCh38

View Full Product Details