Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59191248-59191499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF831
Entrez Gene ID
128611
Gene Name
zinc finger protein 831
Gene Aliases
C20orf174, dJ492J12.1
UniGene
Hs.473204
Gene Chromosome Location
Chr.20: 59146073-59259113 on Build GRCh38
Transcripts
NM_178457.2 XM_005260272.3 XM_005260273.3 XM_006723698.3 XM_011528534.2 XM_011528536.2 XM_011528537.2 XM_011528538.2 XM_011528540.2 XM_017027642.1 XM_017027643.1 XM_017027644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3729758 c.382C>A p.L128M Chr.20: 59191401-59191401 on Build GRCh38
7399752 c.351C>G p.I117M Chr.20: 59191370-59191370 on Build GRCh38
1681803 c.277C>T p.P93S Chr.20: 59191296-59191296 on Build GRCh38
2765070 c.415C>T p.R139W Chr.20: 59191434-59191434 on Build GRCh38
4630600 c.452G>A p.R151H Chr.20: 59191471-59191471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757820738 Chr.20: 59191386-59191386 on Build GRCh38
rs767433729 Chr.20: 59191452-59191452 on Build GRCh38
rs143620250 Chr.20: 59191411-59191411 on Build GRCh38
rs375640811 Chr.20: 59191459-59191459 on Build GRCh38
rs147425916 Chr.20: 59191328-59191328 on Build GRCh38
rs866199440 Chr.20: 59191335-59191335 on Build GRCh38
rs370779472 Chr.20: 59191271-59191271 on Build GRCh38
rs758842616 Chr.20: 59191477-59191477 on Build GRCh38
rs753436459 Chr.20: 59191467-59191467 on Build GRCh38
rs759482756 Chr.20: 59191445-59191445 on Build GRCh38
rs766198515 Chr.20: 59191351-59191351 on Build GRCh38
rs374472847 Chr.20: 59191348-59191348 on Build GRCh38
rs1038816016 Chr.20: 59191367-59191367 on Build GRCh38
rs751452458 Chr.20: 59191354-59191354 on Build GRCh38
rs772454349 Chr.20: 59191399-59191399 on Build GRCh38
rs749815984 Chr.20: 59191317-59191317 on Build GRCh38
rs959081478 Chr.20: 59191316-59191316 on Build GRCh38
rs768759729 Chr.20: 59191402-59191402 on Build GRCh38
rs748743251 Chr.20: 59191379-59191379 on Build GRCh38
rs764029741 Chr.20: 59191266-59191266 on Build GRCh38
rs750931611 Chr.20: 59191475-59191475 on Build GRCh38
rs377419929 Chr.20: 59191391-59191391 on Build GRCh38
rs754843638 Chr.20: 59191360-59191360 on Build GRCh38
rs756739382 Chr.20: 59191470-59191470 on Build GRCh38
rs374647671 Chr.20: 59191305-59191305 on Build GRCh38
rs779368758 Chr.20: 59191393-59191393 on Build GRCh38
rs200287773 Chr.20: 59191412-59191412 on Build GRCh38
rs771328854 Chr.20: 59191321-59191321 on Build GRCh38
rs767382644 Chr.20: 59191361-59191361 on Build GRCh38
rs911835114 Chr.20: 59191345-59191345 on Build GRCh38
rs756321769 Chr.20: 59191394-59191394 on Build GRCh38
rs746198122 Chr.20: 59191325-59191325 on Build GRCh38
rs541763402 Chr.20: 59191314-59191314 on Build GRCh38
rs752403024 Chr.20: 59191364-59191364 on Build GRCh38
rs138229512 Chr.20: 59191400-59191400 on Build GRCh38
rs777399101 Chr.20: 59191371-59191371 on Build GRCh38
rs1015835013 Chr.20: 59191437-59191437 on Build GRCh38
rs763622723 Chr.20: 59191461-59191461 on Build GRCh38
rs751399619 Chr.20: 59191278-59191278 on Build GRCh38
rs748740851 Chr.20: 59191267-59191267 on Build GRCh38
rs371038563 Chr.20: 59191332-59191332 on Build GRCh38
rs912578896 Chr.20: 59191462-59191462 on Build GRCh38
rs7264193 Chr.20: 59191301-59191301 on Build GRCh38
rs760841956 Chr.20: 59191334-59191334 on Build GRCh38
rs780973879 Chr.20: 59191299-59191299 on Build GRCh38
rs746322551 Chr.20: 59191395-59191395 on Build GRCh38
rs750402015 Chr.20: 59191268-59191268 on Build GRCh38
rs563227331 Chr.20: 59191324-59191324 on Build GRCh38
rs778350290 Chr.20: 59191472-59191472 on Build GRCh38
rs535831924 Chr.20: 59191455-59191455 on Build GRCh38
rs200510991 Chr.20: 59191281-59191281 on Build GRCh38
rs774462944 Chr.20: 59191444-59191444 on Build GRCh38
rs202010634 Chr.20: 59191350-59191350 on Build GRCh38
rs761361558 Chr.20: 59191410-59191410 on Build GRCh38
rs758200213 Chr.20: 59191269-59191269 on Build GRCh38
rs750507965 Chr.20: 59191342-59191342 on Build GRCh38
rs528539007 Chr.20: 59191370-59191370 on Build GRCh38
rs201880381 Chr.20: 59191405-59191405 on Build GRCh38

View Full Product Details