Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102944199-102944466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2108563 c.1337G>A p.G446E Chr.11: 102944345-102944345 on Build GRCh38
5371518 c.1400C>T p.S467F Chr.11: 102944282-102944282 on Build GRCh38
4451709 c.1366A>C p.S456R Chr.11: 102944316-102944316 on Build GRCh38
257416 c.1381C>T p.R461C Chr.11: 102944301-102944301 on Build GRCh38
4426372 c.1321A>T p.I441F Chr.11: 102944361-102944361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374512736 Chr.11: 102944299-102944299 on Build GRCh38
rs371369398 Chr.11: 102944385-102944385 on Build GRCh38
rs781986779 Chr.11: 102944392-102944392 on Build GRCh38
rs782669761 Chr.11: 102944222-102944222 on Build GRCh38
rs887296199 Chr.11: 102944281-102944281 on Build GRCh38
rs142064825 Chr.11: 102944310-102944310 on Build GRCh38
rs782182853 Chr.11: 102944325-102944325 on Build GRCh38
rs144776439 Chr.11: 102944305-102944305 on Build GRCh38
rs372513971 Chr.11: 102944253-102944253 on Build GRCh38
rs139939501 Chr.11: 102944346-102944346 on Build GRCh38
rs1022652795 Chr.11: 102944323-102944323 on Build GRCh38
rs782229716 Chr.11: 102944374-102944374 on Build GRCh38
rs145432284 Chr.11: 102944347-102944347 on Build GRCh38
rs375716883 Chr.11: 102944405-102944405 on Build GRCh38
rs782216767 Chr.11: 102944234-102944234 on Build GRCh38
rs370434891 Chr.11: 102944380-102944380 on Build GRCh38
rs978700921 Chr.11: 102944338-102944338 on Build GRCh38
rs782085134 Chr.11: 102944310-102944310 on Build GRCh38
rs782819509 Chr.11: 102944389-102944389 on Build GRCh38
rs782626500 Chr.11: 102944321-102944321 on Build GRCh38
rs782538264 Chr.11: 102944335-102944335 on Build GRCh38
rs782298162 Chr.11: 102944227-102944227 on Build GRCh38
rs781926520 Chr.11: 102944264-102944264 on Build GRCh38
rs781796295 Chr.11: 102944373-102944374 on Build GRCh38
rs944133639 Chr.11: 102944434-102944434 on Build GRCh38
rs368928667 Chr.11: 102944343-102944343 on Build GRCh38
rs781983836 Chr.11: 102944282-102944282 on Build GRCh38
rs782725905 Chr.11: 102944292-102944292 on Build GRCh38
rs782311059 Chr.11: 102944377-102944377 on Build GRCh38
rs781966264 Chr.11: 102944298-102944298 on Build GRCh38
rs782476333 Chr.11: 102944315-102944315 on Build GRCh38
rs558960918 Chr.11: 102944301-102944301 on Build GRCh38
rs956102040 Chr.11: 102944352-102944352 on Build GRCh38
rs782387090 Chr.11: 102944355-102944355 on Build GRCh38
rs782717273 Chr.11: 102944350-102944351 on Build GRCh38
rs782130310 Chr.11: 102944287-102944287 on Build GRCh38
rs781952022 Chr.11: 102944357-102944357 on Build GRCh38

View Full Product Details