Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94997303-94997564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM4D
Entrez Gene ID
55693
Gene Name
lysine demethylase 4D
Gene Aliases
JMJD2D
UniGene
Hs.503598
Gene Chromosome Location
Chr.11: 94973681-94999514 on Build GRCh38
Transcripts
NM_018039.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6519489 c.136G>A p.A46T Chr.11: 94997508-94997508 on Build GRCh38
356706 c.79G>A p.E27K Chr.11: 94997451-94997451 on Build GRCh38
4461523 c.120C>T p.S40S Chr.11: 94997492-94997492 on Build GRCh38
1357981 c.186C>T p.T62T Chr.11: 94997558-94997558 on Build GRCh38
6692491 c.186C>A p.T62T Chr.11: 94997558-94997558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782304288 Chr.11: 94997495-94997495 on Build GRCh38
rs782517456 Chr.11: 94997355-94997355 on Build GRCh38
rs143813885 Chr.11: 94997371-94997371 on Build GRCh38
rs150627695 Chr.11: 94997413-94997413 on Build GRCh38
rs782680440 Chr.11: 94997405-94997405 on Build GRCh38
rs911480841 Chr.11: 94997380-94997380 on Build GRCh38
rs782482345 Chr.11: 94997393-94997393 on Build GRCh38
rs781932328 Chr.11: 94997504-94997504 on Build GRCh38
rs782276207 Chr.11: 94997353-94997353 on Build GRCh38
rs782145835 Chr.11: 94997506-94997506 on Build GRCh38
rs782571259 Chr.11: 94997394-94997394 on Build GRCh38
rs943015494 Chr.11: 94997430-94997430 on Build GRCh38
rs782410456 Chr.11: 94997403-94997403 on Build GRCh38
rs781884920 Chr.11: 94997335-94997335 on Build GRCh38
rs781812597 Chr.11: 94997343-94997343 on Build GRCh38
rs782203256 Chr.11: 94997396-94997396 on Build GRCh38
rs782053255 Chr.11: 94997404-94997404 on Build GRCh38
rs782091808 Chr.11: 94997453-94997453 on Build GRCh38
rs782769893 Chr.11: 94997532-94997532 on Build GRCh38
rs781950501 Chr.11: 94997428-94997428 on Build GRCh38
rs782650005 Chr.11: 94997365-94997365 on Build GRCh38
rs782393734 Chr.11: 94997414-94997414 on Build GRCh38

View Full Product Details