Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 156120705-156120934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB1
Entrez Gene ID
7881
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
Gene Aliases
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
UniGene
Hs.654519
Gene Chromosome Location
Chr.3: 156118211-156540603 on Build GRCh38
Transcripts
NM_001308217.1 NM_172160.2 XM_017007171.1 XM_017007174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4923933 c.179T>A p.L60Q Chr.3: 156120790-156120790 on Build GRCh38
5925055 c.208T>G p.W70G Chr.3: 156120819-156120819 on Build GRCh38
4917160 c.115T>C p.S39P Chr.3: 156120726-156120726 on Build GRCh38
86838 c.189G>T p.L63L Chr.3: 156120800-156120800 on Build GRCh38
7333211 c.214C>T p.L72L Chr.3: 156120825-156120825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780617408 Chr.3: 156120894-156120894 on Build GRCh38
rs768111009 Chr.3: 156120749-156120759 on Build GRCh38
rs778271409 Chr.3: 156120779-156120779 on Build GRCh38
rs1023508537 Chr.3: 156120743-156120743 on Build GRCh38
rs955149217 Chr.3: 156120788-156120788 on Build GRCh38
rs943144521 Chr.3: 156120822-156120822 on Build GRCh38
rs759145411 Chr.3: 156120746-156120746 on Build GRCh38
rs934581727 Chr.3: 156120880-156120880 on Build GRCh38
rs201203895 Chr.3: 156120909-156120909 on Build GRCh38
rs963657391 Chr.3: 156120811-156120811 on Build GRCh38
rs375932233 Chr.3: 156120903-156120903 on Build GRCh38
rs376332854 Chr.3: 156120914-156120914 on Build GRCh38
rs377199610 Chr.3: 156120803-156120803 on Build GRCh38
rs576586282 Chr.3: 156120911-156120911 on Build GRCh38
rs758396951 Chr.3: 156120872-156120872 on Build GRCh38
rs566080944 Chr.3: 156120876-156120876 on Build GRCh38
rs202032013 Chr.3: 156120857-156120857 on Build GRCh38
rs754281331 Chr.3: 156120849-156120849 on Build GRCh38
rs72554022 Chr.3: 156120738-156120738 on Build GRCh38
rs72558049 Chr.3: 156120818-156120818 on Build GRCh38
rs774255657 Chr.3: 156120908-156120908 on Build GRCh38
rs767044176 Chr.3: 156120751-156120751 on Build GRCh38
rs762881758 Chr.3: 156120734-156120734 on Build GRCh38
rs751091184 Chr.3: 156120745-156120745 on Build GRCh38
rs751017939 Chr.3: 156120869-156120869 on Build GRCh38
rs770882880 Chr.3: 156120830-156120830 on Build GRCh38
rs149673348 Chr.3: 156120802-156120802 on Build GRCh38
rs369224487 Chr.3: 156120906-156120906 on Build GRCh38
rs755250066 Chr.3: 156120898-156120898 on Build GRCh38
rs564315619 Chr.3: 156120749-156120749 on Build GRCh38
rs9812862 Chr.3: 156120848-156120848 on Build GRCh38
rs757856229 Chr.3: 156120800-156120800 on Build GRCh38
rs759022544 Chr.3: 156120833-156120833 on Build GRCh38
rs1017119964 Chr.3: 156120778-156120778 on Build GRCh38
rs140686365 Chr.3: 156120782-156120782 on Build GRCh38
rs756418791 Chr.3: 156120774-156120774 on Build GRCh38
rs764168747 Chr.3: 156120892-156120892 on Build GRCh38
rs200450023 Chr.3: 156120831-156120831 on Build GRCh38
rs372851477 Chr.3: 156120781-156120781 on Build GRCh38
rs762827922 Chr.3: 156120826-156120826 on Build GRCh38
rs767109336 Chr.3: 156120841-156120841 on Build GRCh38
rs368292882 Chr.3: 156120810-156120810 on Build GRCh38
rs150921882 Chr.3: 156120757-156120757 on Build GRCh38
rs999686131 Chr.3: 156120741-156120741 on Build GRCh38
rs757799382 Chr.3: 156120850-156120850 on Build GRCh38
rs780117292 Chr.3: 156120804-156120804 on Build GRCh38
rs775004168 Chr.3: 156120846-156120846 on Build GRCh38

View Full Product Details