Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16462663-16462922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOSTDC1
Entrez Gene ID
25928
Gene Name
sclerostin domain containing 1
Gene Aliases
CDA019, DAND7, ECTODIN, USAG1
UniGene
Hs.648106
Gene Chromosome Location
Chr.7: 16461481-16496513 on Build GRCh38
Transcripts
NM_015464.2 XM_017011943.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5976315 c.254C>A p.S85Y Chr.7: 16462915-16462915 on Build GRCh38
5811344 c.326T>C p.V109A Chr.7: 16462843-16462843 on Build GRCh38
6354517 c.306C>T p.G102G Chr.7: 16462863-16462863 on Build GRCh38
1449753 c.355G>T p.G119* Chr.7: 16462814-16462814 on Build GRCh38
6052250 c.448G>A p.G150S Chr.7: 16462721-16462721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759245928 Chr.7: 16462810-16462810 on Build GRCh38
rs767159898 Chr.7: 16462689-16462689 on Build GRCh38
rs770948074 Chr.7: 16462711-16462711 on Build GRCh38
rs868105716 Chr.7: 16462839-16462839 on Build GRCh38
rs964211812 Chr.7: 16462715-16462715 on Build GRCh38
rs761035981 Chr.7: 16462742-16462743 on Build GRCh38
rs778093572 Chr.7: 16462710-16462710 on Build GRCh38
rs778593686 Chr.7: 16462805-16462805 on Build GRCh38
rs143801072 Chr.7: 16462765-16462765 on Build GRCh38
rs367980297 Chr.7: 16462753-16462753 on Build GRCh38
rs774587219 Chr.7: 16462863-16462863 on Build GRCh38
rs917943840 Chr.7: 16462816-16462816 on Build GRCh38
rs774009279 Chr.7: 16462755-16462755 on Build GRCh38
rs761591569 Chr.7: 16462760-16462760 on Build GRCh38
rs1045364308 Chr.7: 16462838-16462838 on Build GRCh38
rs758542305 Chr.7: 16462708-16462708 on Build GRCh38
rs140346889 Chr.7: 16462698-16462698 on Build GRCh38
rs147807617 Chr.7: 16462731-16462731 on Build GRCh38
rs749784692 Chr.7: 16462853-16462853 on Build GRCh38
rs761503585 Chr.7: 16462889-16462889 on Build GRCh38
rs200731488 Chr.7: 16462783-16462783 on Build GRCh38
rs746015616 Chr.7: 16462844-16462844 on Build GRCh38
rs749970369 Chr.7: 16462767-16462767 on Build GRCh38
rs374376320 Chr.7: 16462819-16462819 on Build GRCh38
rs766402142 Chr.7: 16462772-16462772 on Build GRCh38
rs780662318 Chr.7: 16462850-16462850 on Build GRCh38
rs754848341 Chr.7: 16462788-16462788 on Build GRCh38
rs770345144 Chr.7: 16462737-16462737 on Build GRCh38
rs769028816 Chr.7: 16462854-16462854 on Build GRCh38
rs750350294 Chr.7: 16462704-16462704 on Build GRCh38
rs540348040 Chr.7: 16462828-16462828 on Build GRCh38
rs760672874 Chr.7: 16462771-16462771 on Build GRCh38
rs115190299 Chr.7: 16462707-16462707 on Build GRCh38
rs770171919 Chr.7: 16462847-16462847 on Build GRCh38
rs370368434 Chr.7: 16462754-16462754 on Build GRCh38
rs761461062 Chr.7: 16462686-16462686 on Build GRCh38
rs34300902 Chr.7: 16462821-16462821 on Build GRCh38
rs781280784 Chr.7: 16462716-16462716 on Build GRCh38
rs1001089128 Chr.7: 16462862-16462862 on Build GRCh38
rs771904504 Chr.7: 16462899-16462899 on Build GRCh38
rs111420154 Chr.7: 16462785-16462785 on Build GRCh38
rs35600737 Chr.7: 16462756-16462757 on Build GRCh38

View Full Product Details