Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53498302-53498563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
UniGene
Hs.380897
Gene Chromosome Location
Chr.16: 53491028-53504411 on Build GRCh38
Transcripts
NM_001012398.2 NM_001308325.1 NM_022476.3 XM_005256095.4 XM_005256096.4 XM_005256097.4 XM_005256098.4 XM_017023564.1 XM_017023565.1 XM_017023566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660483 c.133A>G p.K45E Chr.16: 53498506-53498506 on Build GRCh38
7185639 c.228delA p.E76fs*12 Chr.16: 53498411-53498411 on Build GRCh38
6144842 c.130C>T p.P44S Chr.16: 53498509-53498509 on Build GRCh38
6481553 c.97C>T p.R33* Chr.16: 53498542-53498542 on Build GRCh38
3932365 c.151A>G p.T51A Chr.16: 53498488-53498488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746359591 Chr.16: 53498424-53498424 on Build GRCh38
rs754564038 Chr.16: 53498435-53498435 on Build GRCh38
rs750978594 Chr.16: 53498501-53498501 on Build GRCh38
rs758521799 Chr.16: 53498372-53498372 on Build GRCh38
rs1021665141 Chr.16: 53498452-53498452 on Build GRCh38
rs747653033 Chr.16: 53498441-53498441 on Build GRCh38
rs893655997 Chr.16: 53498346-53498346 on Build GRCh38
rs192175938 Chr.16: 53498332-53498332 on Build GRCh38
rs759730470 Chr.16: 53498345-53498345 on Build GRCh38
rs757561778 Chr.16: 53498387-53498387 on Build GRCh38
rs765221835 Chr.16: 53498361-53498361 on Build GRCh38
rs767844319 Chr.16: 53498491-53498491 on Build GRCh38
rs746802147 Chr.16: 53498449-53498449 on Build GRCh38
rs759373692 Chr.16: 53498467-53498467 on Build GRCh38
rs770843306 Chr.16: 53498460-53498460 on Build GRCh38
rs1038130700 Chr.16: 53498471-53498471 on Build GRCh38
rs751944152 Chr.16: 53498378-53498378 on Build GRCh38
rs756609255 Chr.16: 53498514-53498514 on Build GRCh38
rs751715236 Chr.16: 53498481-53498481 on Build GRCh38
rs776770248 Chr.16: 53498465-53498465 on Build GRCh38
rs199548307 Chr.16: 53498475-53498475 on Build GRCh38
rs199669647 Chr.16: 53498488-53498488 on Build GRCh38
rs750826158 Chr.16: 53498471-53498479 on Build GRCh38
rs1046908213 Chr.16: 53498454-53498454 on Build GRCh38
rs775464421 Chr.16: 53498473-53498473 on Build GRCh38
rs778254783 Chr.16: 53498436-53498436 on Build GRCh38
rs764261874 Chr.16: 53498376-53498376 on Build GRCh38
rs753017265 Chr.16: 53498367-53498367 on Build GRCh38
rs908389472 Chr.16: 53498455-53498455 on Build GRCh38
rs141560947 Chr.16: 53498541-53498541 on Build GRCh38
rs763039734 Chr.16: 53498477-53498477 on Build GRCh38
rs772937741 Chr.16: 53498444-53498444 on Build GRCh38
rs771641760 Chr.16: 53498442-53498442 on Build GRCh38
rs765801475 Chr.16: 53498375-53498375 on Build GRCh38
rs533946422 Chr.16: 53498482-53498482 on Build GRCh38
rs903240268 Chr.16: 53498453-53498453 on Build GRCh38
rs986386188 Chr.16: 53498511-53498511 on Build GRCh38
rs139608589 Chr.16: 53498411-53498411 on Build GRCh38
rs757855483 Chr.16: 53498537-53498537 on Build GRCh38
rs747621138 Chr.16: 53498530-53498530 on Build GRCh38
rs780558074 Chr.16: 53498528-53498528 on Build GRCh38
rs764315195 Chr.16: 53498478-53498478 on Build GRCh38

View Full Product Details