Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15737918-15738191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A34
Entrez Gene ID
284723
Gene Name
solute carrier family 25 member 34
Gene Aliases
-
UniGene
Hs.631867
Gene Chromosome Location
Chr.1: 15735088-15741392 on Build GRCh38
Transcripts
NM_207348.2 XM_011541292.2 XM_011541293.1 XM_017001083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4024224 c.475C>T p.R159W Chr.1: 15738123-15738123 on Build GRCh38
6037065 c.391C>T p.L131L Chr.1: 15737941-15737941 on Build GRCh38
898341 c.507C>T p.G169G Chr.1: 15738155-15738155 on Build GRCh38
4024223 c.432G>A p.Q144Q Chr.1: 15737982-15737982 on Build GRCh38
7390289 c.421G>A p.V141M Chr.1: 15737971-15737971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754136678 Chr.1: 15738017-15738017 on Build GRCh38
rs202068853 Chr.1: 15738065-15738065 on Build GRCh38
rs762363289 Chr.1: 15737991-15737991 on Build GRCh38
rs199643953 Chr.1: 15738055-15738055 on Build GRCh38
rs759405576 Chr.1: 15737983-15737983 on Build GRCh38
rs769775687 Chr.1: 15738061-15738061 on Build GRCh38
rs779847919 Chr.1: 15738163-15738163 on Build GRCh38
rs58307910 Chr.1: 15737955-15737955 on Build GRCh38
rs769452605 Chr.1: 15738156-15738156 on Build GRCh38
rs756564600 Chr.1: 15738008-15738008 on Build GRCh38
rs139368754 Chr.1: 15738123-15738123 on Build GRCh38
rs370099080 Chr.1: 15738066-15738066 on Build GRCh38
rs751002802 Chr.1: 15737949-15737949 on Build GRCh38
rs770831428 Chr.1: 15737972-15737972 on Build GRCh38
rs755350415 Chr.1: 15738019-15738019 on Build GRCh38
rs200358223 Chr.1: 15737961-15737961 on Build GRCh38
rs768998208 Chr.1: 15738057-15738057 on Build GRCh38
rs377690904 Chr.1: 15738075-15738075 on Build GRCh38
rs747009687 Chr.1: 15737971-15737971 on Build GRCh38
rs770955830 Chr.1: 15738048-15738048 on Build GRCh38
rs992844330 Chr.1: 15738059-15738059 on Build GRCh38
rs369422240 Chr.1: 15738001-15738001 on Build GRCh38
rs371293754 Chr.1: 15738167-15738167 on Build GRCh38
rs549032562 Chr.1: 15738027-15738027 on Build GRCh38
rs781285161 Chr.1: 15738049-15738049 on Build GRCh38
rs376238856 Chr.1: 15737954-15737954 on Build GRCh38
rs747112560 Chr.1: 15738043-15738043 on Build GRCh38
rs777960646 Chr.1: 15738039-15738039 on Build GRCh38
rs777892152 Chr.1: 15738129-15738129 on Build GRCh38
rs372996692 Chr.1: 15738024-15738024 on Build GRCh38
rs146057979 Chr.1: 15737962-15737962 on Build GRCh38
rs763698841 Chr.1: 15738000-15738000 on Build GRCh38
rs1030690025 Chr.1: 15738158-15738158 on Build GRCh38
rs749675323 Chr.1: 15737957-15737957 on Build GRCh38
rs749352608 Chr.1: 15738137-15738137 on Build GRCh38
rs772253470 Chr.1: 15737967-15737967 on Build GRCh38
rs766901791 Chr.1: 15738016-15738016 on Build GRCh38
rs745838193 Chr.1: 15738152-15738152 on Build GRCh38
rs575582816 Chr.1: 15738116-15738116 on Build GRCh38
rs776523075 Chr.1: 15737979-15737979 on Build GRCh38
rs759891454 Chr.1: 15738119-15738119 on Build GRCh38
rs916539702 Chr.1: 15738140-15738140 on Build GRCh38
rs773871259 Chr.1: 15738076-15738076 on Build GRCh38
rs766814031 Chr.1: 15738114-15738114 on Build GRCh38
rs900436615 Chr.1: 15737948-15737948 on Build GRCh38
rs12025441 Chr.1: 15737982-15737982 on Build GRCh38
rs974241177 Chr.1: 15738029-15738029 on Build GRCh38
rs768286992 Chr.1: 15738174-15738174 on Build GRCh38
rs571515007 Chr.1: 15738155-15738155 on Build GRCh38
rs768789504 Chr.1: 15738150-15738150 on Build GRCh38
rs1016031881 Chr.1: 15737973-15737973 on Build GRCh38
rs151316306 Chr.1: 15737970-15737970 on Build GRCh38
rs200722262 Chr.1: 15738143-15738143 on Build GRCh38
rs373094971 Chr.1: 15738053-15738053 on Build GRCh38
rs867736253 Chr.1: 15738142-15738142 on Build GRCh38
rs201053751 Chr.1: 15737946-15737946 on Build GRCh38
rs781268915 Chr.1: 15738151-15738151 on Build GRCh38
rs765674952 Chr.1: 15738120-15738120 on Build GRCh38
rs764920680 Chr.1: 15737986-15737986 on Build GRCh38
rs926742696 Chr.1: 15738040-15738040 on Build GRCh38
rs758602423 Chr.1: 15738124-15738124 on Build GRCh38
rs761316078 Chr.1: 15738096-15738096 on Build GRCh38

View Full Product Details