Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 44397155-44397398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERP2
Entrez Gene ID
387923
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Gene Aliases
C13orf21, bA269C23.1
UniGene
Hs.377972
Gene Chromosome Location
Chr.13: 44373842-44397714 on Build GRCh38
Transcripts
NM_001010897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6775455 c.171C>T p.I57I Chr.13: 44397285-44397285 on Build GRCh38
4443260 c.169A>G p.I57V Chr.13: 44397283-44397283 on Build GRCh38
6576546 c.166C>T p.Q56* Chr.13: 44397280-44397280 on Build GRCh38
1747452 c.178A>G p.S60G Chr.13: 44397292-44397292 on Build GRCh38
6868844 c.195G>A p.M65I Chr.13: 44397309-44397309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372132224 Chr.13: 44397355-44397355 on Build GRCh38
rs779011597 Chr.13: 44397325-44397325 on Build GRCh38
rs757516505 Chr.13: 44397251-44397253 on Build GRCh38
rs2296272 Chr.13: 44397207-44397207 on Build GRCh38
rs761321699 Chr.13: 44397339-44397339 on Build GRCh38
rs748201523 Chr.13: 44397335-44397335 on Build GRCh38
rs41288337 Chr.13: 44397367-44397367 on Build GRCh38
rs538946533 Chr.13: 44397234-44397234 on Build GRCh38
rs774308237 Chr.13: 44397226-44397226 on Build GRCh38
rs887354340 Chr.13: 44397186-44397186 on Build GRCh38
rs368745065 Chr.13: 44397348-44397348 on Build GRCh38
rs759123816 Chr.13: 44397265-44397265 on Build GRCh38
rs1047296300 Chr.13: 44397177-44397177 on Build GRCh38
rs772231150 Chr.13: 44397336-44397336 on Build GRCh38
rs764816852 Chr.13: 44397364-44397364 on Build GRCh38
rs779318430 Chr.13: 44397280-44397282 on Build GRCh38
rs777136385 Chr.13: 44397354-44397354 on Build GRCh38
rs768765909 Chr.13: 44397300-44397300 on Build GRCh38
rs1000567394 Chr.13: 44397365-44397365 on Build GRCh38
rs781026727 Chr.13: 44397294-44397294 on Build GRCh38
rs372274801 Chr.13: 44397230-44397230 on Build GRCh38
rs74508112 Chr.13: 44397252-44397252 on Build GRCh38
rs773849864 Chr.13: 44397338-44397338 on Build GRCh38
rs749403196 Chr.13: 44397245-44397246 on Build GRCh38
rs571308098 Chr.13: 44397210-44397210 on Build GRCh38
rs373364072 Chr.13: 44397211-44397211 on Build GRCh38
rs900086939 Chr.13: 44397371-44397371 on Build GRCh38
rs773664008 Chr.13: 44397240-44397240 on Build GRCh38
rs766916319 Chr.13: 44397257-44397257 on Build GRCh38
rs749469457 Chr.13: 44397296-44397296 on Build GRCh38
rs917113582 Chr.13: 44397319-44397319 on Build GRCh38
rs963344946 Chr.13: 44397179-44397179 on Build GRCh38
rs904615663 Chr.13: 44397333-44397333 on Build GRCh38
rs753526770 Chr.13: 44397261-44397261 on Build GRCh38
rs868674987 Chr.13: 44397205-44397205 on Build GRCh38
rs758303272 Chr.13: 44397272-44397272 on Build GRCh38
rs1042168701 Chr.13: 44397332-44397332 on Build GRCh38
rs751540932 Chr.13: 44397285-44397285 on Build GRCh38
rs764733985 Chr.13: 44397268-44397268 on Build GRCh38
rs746787500 Chr.13: 44397317-44397318 on Build GRCh38
rs537931253 Chr.13: 44397184-44397184 on Build GRCh38
rs777662445 Chr.13: 44397274-44397274 on Build GRCh38
rs757044571 Chr.13: 44397289-44397289 on Build GRCh38
rs551491504 Chr.13: 44397323-44397323 on Build GRCh38
rs567418338 Chr.13: 44397175-44397175 on Build GRCh38

View Full Product Details