Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 40730139-40730388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGER4
Entrez Gene ID
5734
Gene Name
prostaglandin E receptor 4
Gene Aliases
EP4, EP4R
UniGene
Hs.199248
Gene Chromosome Location
Chr.5: 40679930-40740932 on Build GRCh38
Transcripts
XM_017009659.1 NM_012382.2 XM_011514003.2

Gene Symbol
TTC33
Entrez Gene ID
23548
Gene Name
tetratricopeptide repeat domain 33
Gene Aliases
OSRF
UniGene
Hs.348915
Gene Chromosome Location
Chr.5: 40711576-40756965 on Build GRCh38
Transcripts
XM_017009659.1 NM_012382.2 XM_011514003.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5868529 c.226C>T p.R76W Chr.5: 40730339-40730339 on Build GRCh38
420699 c.294G>A p.M98I Chr.5: 40730271-40730271 on Build GRCh38
3702815 c.279T>C p.A93A Chr.5: 40730286-40730286 on Build GRCh38
6406985 c.283C>A p.L95I Chr.5: 40730282-40730282 on Build GRCh38
7262613 c.288C>T p.Y96Y Chr.5: 40730277-40730277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755328365 Chr.5: 40730340-40730340 on Build GRCh38
rs375359844 Chr.5: 40730250-40730250 on Build GRCh38
rs760892150 Chr.5: 40730208-40730208 on Build GRCh38
rs779286125 Chr.5: 40730353-40730353 on Build GRCh38
rs191767399 Chr.5: 40730223-40730223 on Build GRCh38
rs374276458 Chr.5: 40730338-40730338 on Build GRCh38
rs775272772 Chr.5: 40730241-40730241 on Build GRCh38
rs767421056 Chr.5: 40730311-40730311 on Build GRCh38
rs369422414 Chr.5: 40730325-40730325 on Build GRCh38
rs537565933 Chr.5: 40730209-40730209 on Build GRCh38
rs757720483 Chr.5: 40730217-40730218 on Build GRCh38
rs1009260811 Chr.5: 40730317-40730317 on Build GRCh38
rs144481517 Chr.5: 40730276-40730276 on Build GRCh38
rs765624558 Chr.5: 40730224-40730224 on Build GRCh38
rs201413319 Chr.5: 40730337-40730337 on Build GRCh38
rs763885705 Chr.5: 40730251-40730251 on Build GRCh38
rs761762250 Chr.5: 40730296-40730296 on Build GRCh38
rs751422389 Chr.5: 40730339-40730339 on Build GRCh38
rs917567154 Chr.5: 40730169-40730169 on Build GRCh38

View Full Product Details