Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101461900-101462131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTRC
Entrez Gene ID
8945
Gene Name
beta-transducin repeat containing E3 ubiquitin protein ligase
Gene Aliases
BETA-TRCP, FBW1A, FBXW1, FBXW1A, FWD1, bTrCP, bTrCP1, betaTrCP
UniGene
Hs.643802
Gene Chromosome Location
Chr.10: 101353808-101557321 on Build GRCh38
Transcripts
NM_001256856.1 NM_003939.4 NM_033637.3 XM_006718054.2 XM_017016870.1 XM_017016871.1 XM_017016872.1 XM_017016873.1 XM_017016874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6211948 c.179G>T p.C60F Chr.10: 101462003-101462003 on Build GRCh38
4011121 c.189C>T p.G63G Chr.10: 101462013-101462013 on Build GRCh38
6128194 c.164C>A p.S55* Chr.10: 101461988-101461988 on Build GRCh38
7303735 c.212C>G p.P71R Chr.10: 101462036-101462036 on Build GRCh38
73873 c.196C>T p.P66S Chr.10: 101462020-101462020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762652665 Chr.10: 101462077-101462077 on Build GRCh38
rs775187144 Chr.10: 101462053-101462056 on Build GRCh38
rs139681293 Chr.10: 101461946-101461946 on Build GRCh38
rs767784372 Chr.10: 101462079-101462083 on Build GRCh38
rs572930830 Chr.10: 101461994-101461994 on Build GRCh38
rs746001139 Chr.10: 101462107-101462107 on Build GRCh38
rs757945127 Chr.10: 101462072-101462072 on Build GRCh38
rs759103744 Chr.10: 101462013-101462013 on Build GRCh38
rs368554823 Chr.10: 101461936-101461936 on Build GRCh38
rs558190836 Chr.10: 101461953-101461953 on Build GRCh38
rs765030761 Chr.10: 101462014-101462014 on Build GRCh38
rs746995431 Chr.10: 101461933-101461933 on Build GRCh38
rs752361392 Chr.10: 101462037-101462037 on Build GRCh38
rs569138071 Chr.10: 101461962-101461962 on Build GRCh38
rs777643075 Chr.10: 101462079-101462079 on Build GRCh38
rs371336032 Chr.10: 101461948-101461948 on Build GRCh38
rs763270900 Chr.10: 101461964-101461964 on Build GRCh38
rs769098517 Chr.10: 101461967-101461967 on Build GRCh38
rs1046551524 Chr.10: 101462059-101462059 on Build GRCh38
rs949189845 Chr.10: 101462056-101462056 on Build GRCh38
rs751373171 Chr.10: 101461937-101461937 on Build GRCh38
rs368161919 Chr.10: 101462010-101462010 on Build GRCh38
rs768071719 Chr.10: 101462003-101462003 on Build GRCh38
rs745877690 Chr.10: 101461942-101461942 on Build GRCh38
rs374659154 Chr.10: 101461969-101461969 on Build GRCh38
rs751251822 Chr.10: 101462080-101462080 on Build GRCh38
rs774532059 Chr.10: 101461993-101461993 on Build GRCh38
rs757187514 Chr.10: 101462095-101462095 on Build GRCh38
rs1053267536 Chr.10: 101461957-101461957 on Build GRCh38

View Full Product Details