Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128574610-128574883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPTAN1
Entrez Gene ID
6709
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
EIEE5, NEAS, SPTA2
UniGene
Hs.372331
Gene Chromosome Location
Chr.9: 128552558-128633665 on Build GRCh38
Transcripts
NM_001130438.2 NM_001195532.1 NM_003127.3 XM_006717245.1 XM_006717246.1 XM_006717247.2 XM_006717248.2 XM_006717249.1 XM_006717250.1 XM_006717251.2 XM_006717252.2 XM_006717253.1 XM_006717254.2 XM_017015059.1 XM_017015060.1 XM_017015061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6489862 c.497A>T p.N166I Chr.9: 128574808-128574808 on Build GRCh38
3654668 c.444C>T p.A148A Chr.9: 128574755-128574755 on Build GRCh38
3654667 c.444C>T p.A148A Chr.9: 128574755-128574755 on Build GRCh38
4882582 c.481G>T p.V161L Chr.9: 128574792-128574792 on Build GRCh38
752616 c.478G>T p.D160Y Chr.9: 128574789-128574789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748252690 Chr.9: 128574845-128574845 on Build GRCh38
rs773485341 Chr.9: 128574859-128574859 on Build GRCh38
rs377258326 Chr.9: 128574820-128574820 on Build GRCh38
rs139943747 Chr.9: 128574808-128574808 on Build GRCh38
rs749280573 Chr.9: 128574825-128574825 on Build GRCh38
rs780867742 Chr.9: 128574655-128574655 on Build GRCh38
rs747453186 Chr.9: 128574775-128574776 on Build GRCh38
rs746712451 Chr.9: 128574649-128574649 on Build GRCh38
rs776493422 Chr.9: 128574757-128574757 on Build GRCh38
rs754419719 Chr.9: 128574744-128574744 on Build GRCh38
rs775634580 Chr.9: 128574679-128574679 on Build GRCh38
rs1017406868 Chr.9: 128574633-128574633 on Build GRCh38
rs757364107 Chr.9: 128574651-128574651 on Build GRCh38
rs773288631 Chr.9: 128574727-128574727 on Build GRCh38
rs1005495879 Chr.9: 128574839-128574839 on Build GRCh38
rs201997683 Chr.9: 128574860-128574860 on Build GRCh38
rs767527240 Chr.9: 128574724-128574724 on Build GRCh38
rs771370468 Chr.9: 128574853-128574856 on Build GRCh38
rs771009235 Chr.9: 128574693-128574693 on Build GRCh38
rs774397953 Chr.9: 128574697-128574697 on Build GRCh38
rs181402160 Chr.9: 128574690-128574690 on Build GRCh38
rs201249013 Chr.9: 128574791-128574791 on Build GRCh38
rs770303060 Chr.9: 128574650-128574650 on Build GRCh38
rs756980032 Chr.9: 128574800-128574800 on Build GRCh38
rs755742062 Chr.9: 128574819-128574819 on Build GRCh38
rs755431637 Chr.9: 128574645-128574645 on Build GRCh38
rs964919191 Chr.9: 128574696-128574696 on Build GRCh38
rs147900000 Chr.9: 128574715-128574715 on Build GRCh38
rs766860881 Chr.9: 128574737-128574737 on Build GRCh38
rs975951020 Chr.9: 128574701-128574701 on Build GRCh38
rs201763197 Chr.9: 128574639-128574639 on Build GRCh38
rs373503519 Chr.9: 128574647-128574647 on Build GRCh38
rs769857503 Chr.9: 128574678-128574678 on Build GRCh38
rs142120579 Chr.9: 128574702-128574702 on Build GRCh38
rs768538310 Chr.9: 128574691-128574691 on Build GRCh38
rs761107143 Chr.9: 128574770-128574770 on Build GRCh38
rs768287672 Chr.9: 128574835-128574835 on Build GRCh38
rs774209726 Chr.9: 128574836-128574836 on Build GRCh38
rs780782090 Chr.9: 128574804-128574804 on Build GRCh38
rs772281155 Chr.9: 128574857-128574857 on Build GRCh38
rs200978151 Chr.9: 128574656-128574656 on Build GRCh38
rs760815035 Chr.9: 128574734-128574734 on Build GRCh38
rs765662528 Chr.9: 128574784-128574784 on Build GRCh38

View Full Product Details