Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15243036-15243295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD4
Entrez Gene ID
23476
Gene Name
bromodomain containing 4
Gene Aliases
CAP, HUNK1, HUNKI, MCAP
UniGene
Hs.187763
Gene Chromosome Location
Chr.19: 15236836-15332543 on Build GRCh38
Transcripts
NM_058243.2 XM_011527854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403497 c.2784G>T p.M928I Chr.19: 15243285-15243285 on Build GRCh38
6959794 c.2855C>A p.S952Y Chr.19: 15243214-15243214 on Build GRCh38
6933679 c.2842G>A p.V948M Chr.19: 15243227-15243227 on Build GRCh38
6967206 c.2800_2808delCAGCTGCAG p.Q934_Q936delQLQ Chr.19: 15243261-15243269 on Build GRCh38
6950642 c.3014C>T p.S1005F Chr.19: 15243055-15243055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777234614 Chr.19: 15243147-15243147 on Build GRCh38
rs925612648 Chr.19: 15243187-15243201 on Build GRCh38
rs753808683 Chr.19: 15243068-15243068 on Build GRCh38
rs1056025059 Chr.19: 15243077-15243077 on Build GRCh38
rs759439799 Chr.19: 15243243-15243243 on Build GRCh38
rs547971458 Chr.19: 15243093-15243093 on Build GRCh38
rs777912948 Chr.19: 15243086-15243086 on Build GRCh38
rs199555361 Chr.19: 15243226-15243226 on Build GRCh38
rs867317090 Chr.19: 15243110-15243110 on Build GRCh38
rs985647849 Chr.19: 15243267-15243267 on Build GRCh38
rs1030768906 Chr.19: 15243192-15243192 on Build GRCh38
rs528231191 Chr.19: 15243071-15243071 on Build GRCh38
rs777645654 Chr.19: 15243145-15243146 on Build GRCh38
rs955003930 Chr.19: 15243196-15243196 on Build GRCh38
rs866969097 Chr.19: 15243109-15243109 on Build GRCh38
rs767143670 Chr.19: 15243225-15243225 on Build GRCh38
rs1041579791 Chr.19: 15243206-15243206 on Build GRCh38
rs760142472 Chr.19: 15243228-15243228 on Build GRCh38
rs1022195624 Chr.19: 15243141-15243141 on Build GRCh38
rs550633083 Chr.19: 15243123-15243123 on Build GRCh38
rs746959252 Chr.19: 15243182-15243199 on Build GRCh38
rs770471285 Chr.19: 15243200-15243200 on Build GRCh38
rs374518582 Chr.19: 15243258-15243258 on Build GRCh38
rs747935780 Chr.19: 15243080-15243080 on Build GRCh38
rs371213651 Chr.19: 15243240-15243240 on Build GRCh38
rs911446850 Chr.19: 15243152-15243160 on Build GRCh38
rs530810477 Chr.19: 15243121-15243121 on Build GRCh38
rs967600500 Chr.19: 15243181-15243181 on Build GRCh38
rs866351525 Chr.19: 15243118-15243118 on Build GRCh38
rs999654429 Chr.19: 15243186-15243186 on Build GRCh38
rs758265124 Chr.19: 15243128-15243136 on Build GRCh38
rs765227557 Chr.19: 15243244-15243244 on Build GRCh38
rs1011877373 Chr.19: 15243132-15243132 on Build GRCh38
rs561779284 Chr.19: 15243099-15243099 on Build GRCh38
rs1027539682 Chr.19: 15243075-15243075 on Build GRCh38
rs778412067 Chr.19: 15243066-15243080 on Build GRCh38
rs753396296 Chr.19: 15243241-15243241 on Build GRCh38
rs945737077 Chr.19: 15243198-15243198 on Build GRCh38
rs758643407 Chr.19: 15243081-15243081 on Build GRCh38
rs748014251 Chr.19: 15243115-15243116 on Build GRCh38
rs201742792 Chr.19: 15243227-15243227 on Build GRCh38
rs987111459 Chr.19: 15243149-15243160 on Build GRCh38
rs746395414 Chr.19: 15243156-15243156 on Build GRCh38
rs758139232 Chr.19: 15243275-15243275 on Build GRCh38
rs925130615 Chr.19: 15243252-15243252 on Build GRCh38
rs570787628 Chr.19: 15243213-15243213 on Build GRCh38
rs963778083 Chr.19: 15243179-15243179 on Build GRCh38
rs775889378 Chr.19: 15243204-15243204 on Build GRCh38
rs914340499 Chr.19: 15243197-15243197 on Build GRCh38

View Full Product Details