Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75621541-75621772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SENP6
Entrez Gene ID
26054
Gene Name
SUMO1/sentrin specific peptidase 6
Gene Aliases
SSP1, SUSP1
UniGene
Hs.485784
Gene Chromosome Location
Chr.6: 75601509-75718285 on Build GRCh38
Transcripts
NM_001100409.2 NM_001304792.1 NM_015571.3 XM_006715457.3 XM_011535746.2 XM_011535747.2 XM_017010719.1 XM_017010720.1 XM_017010722.1 XM_017010723.1 XM_017010724.1 XM_017010725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556144 c.67G>T p.E23* Chr.6: 75621546-75621546 on Build GRCh38
1643140 c.146+8delT p.? Chr.6: 75621633-75621633 on Build GRCh38
1181025 c.146+8delT p.? Chr.6: 75621633-75621633 on Build GRCh38
4596432 c.122A>G p.E41G Chr.6: 75621601-75621601 on Build GRCh38
5503139 c.146+6G>T p.? Chr.6: 75621631-75621631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34679709 Chr.6: 75621648-75621649 on Build GRCh38
rs943757029 Chr.6: 75621725-75621725 on Build GRCh38
rs368137519 Chr.6: 75621614-75621614 on Build GRCh38
rs757442596 Chr.6: 75621644-75621644 on Build GRCh38
rs751298967 Chr.6: 75621650-75621650 on Build GRCh38
rs756150201 Chr.6: 75621667-75621667 on Build GRCh38
rs891897864 Chr.6: 75621696-75621696 on Build GRCh38
rs757929357 Chr.6: 75621611-75621616 on Build GRCh38
rs745508415 Chr.6: 75621676-75621676 on Build GRCh38
rs368441950 Chr.6: 75621671-75621671 on Build GRCh38
rs1054919077 Chr.6: 75621646-75621646 on Build GRCh38
rs748067954 Chr.6: 75621665-75621665 on Build GRCh38
rs1049261563 Chr.6: 75621743-75621746 on Build GRCh38
rs752274690 Chr.6: 75621574-75621575 on Build GRCh38
rs770745509 Chr.6: 75621601-75621601 on Build GRCh38
rs781251360 Chr.6: 75621571-75621571 on Build GRCh38
rs912232087 Chr.6: 75621693-75621693 on Build GRCh38
rs774209210 Chr.6: 75621675-75621675 on Build GRCh38
rs375689983 Chr.6: 75621649-75621649 on Build GRCh38
rs1034237424 Chr.6: 75621743-75621743 on Build GRCh38
rs777386460 Chr.6: 75621670-75621670 on Build GRCh38
rs753952445 Chr.6: 75621642-75621642 on Build GRCh38
rs746955585 Chr.6: 75621632-75621633 on Build GRCh38
rs774833035 Chr.6: 75621604-75621604 on Build GRCh38
rs765493796 Chr.6: 75621647-75621647 on Build GRCh38
rs772420900 Chr.6: 75621677-75621677 on Build GRCh38
rs1010635556 Chr.6: 75621719-75621719 on Build GRCh38
rs569411536 Chr.6: 75621692-75621692 on Build GRCh38
rs748909335 Chr.6: 75621599-75621599 on Build GRCh38
rs3831261 Chr.6: 75621742-75621743 on Build GRCh38
rs770799162 Chr.6: 75621634-75621644 on Build GRCh38
rs370825373 Chr.6: 75621619-75621619 on Build GRCh38
rs559553859 Chr.6: 75621591-75621591 on Build GRCh38
rs1024290455 Chr.6: 75621731-75621731 on Build GRCh38
rs755166746 Chr.6: 75621736-75621737 on Build GRCh38
rs533363441 Chr.6: 75621641-75621641 on Build GRCh38
rs745743761 Chr.6: 75621648-75621651 on Build GRCh38
rs769452793 Chr.6: 75621654-75621660 on Build GRCh38
rs754870821 Chr.6: 75621656-75621656 on Build GRCh38
rs373949306 Chr.6: 75621612-75621612 on Build GRCh38
rs573890257 Chr.6: 75621633-75621633 on Build GRCh38
rs549001829 Chr.6: 75621739-75621739 on Build GRCh38
rs772871460 Chr.6: 75621596-75621596 on Build GRCh38
rs34451237 Chr.6: 75621592-75621592 on Build GRCh38
rs181706733 Chr.6: 75621664-75621664 on Build GRCh38
rs536705084 Chr.6: 75621738-75621738 on Build GRCh38
rs761102583 Chr.6: 75621621-75621621 on Build GRCh38
rs780978751 Chr.6: 75621634-75621635 on Build GRCh38

View Full Product Details