Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 104787851-104788120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA1
Entrez Gene ID
19
Gene Name
ATP binding cassette subfamily A member 1
Gene Aliases
ABC-1, ABC1, CERP, HDLDT1, TGD
UniGene
Hs.659274
Gene Chromosome Location
Chr.9: 104781002-104928246 on Build GRCh38
Transcripts
NM_005502.3 XM_005251773.2 XM_005251776.2 XM_011518339.2 XM_011518340.2 XM_011518341.2 XM_011518342.2 XM_017014378.1 XM_017014379.1 XM_017014380.1 XM_017014381.1 XM_017014382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836848 c.6088C>T p.R2030W Chr.9: 104788036-104788036 on Build GRCh38
3167951 c.6094C>G p.L2032V Chr.9: 104788030-104788030 on Build GRCh38
6182138 c.6132C>T p.N2044N Chr.9: 104787992-104787992 on Build GRCh38
6114487 c.6118A>C p.K2040Q Chr.9: 104788006-104788006 on Build GRCh38
1243857 c.6084G>A p.A2028A Chr.9: 104788040-104788040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758292384 Chr.9: 104787977-104787977 on Build GRCh38
rs750728670 Chr.9: 104788057-104788057 on Build GRCh38
rs756936070 Chr.9: 104788079-104788079 on Build GRCh38
rs757969860 Chr.9: 104788093-104788093 on Build GRCh38
rs763124062 Chr.9: 104787916-104787916 on Build GRCh38
rs780091906 Chr.9: 104787981-104787981 on Build GRCh38
rs753024806 Chr.9: 104787954-104787954 on Build GRCh38
rs776103826 Chr.9: 104788055-104788055 on Build GRCh38
rs368951069 Chr.9: 104788030-104788030 on Build GRCh38
rs760942503 Chr.9: 104788059-104788059 on Build GRCh38
rs141032263 Chr.9: 104787944-104787944 on Build GRCh38
rs144323320 Chr.9: 104787913-104787913 on Build GRCh38
rs947401662 Chr.9: 104787920-104787920 on Build GRCh38
rs767844796 Chr.9: 104787916-104787916 on Build GRCh38
rs771064359 Chr.9: 104788040-104788040 on Build GRCh38
rs751548064 Chr.9: 104787943-104787943 on Build GRCh38
rs769888806 Chr.9: 104787903-104787903 on Build GRCh38
rs764680261 Chr.9: 104788090-104788090 on Build GRCh38
rs752051518 Chr.9: 104787940-104787940 on Build GRCh38
rs372790037 Chr.9: 104788021-104788021 on Build GRCh38
rs762048296 Chr.9: 104787892-104787892 on Build GRCh38
rs753554858 Chr.9: 104787963-104787963 on Build GRCh38
rs953062048 Chr.9: 104788073-104788073 on Build GRCh38
rs1001267024 Chr.9: 104787967-104787967 on Build GRCh38
rs762842594 Chr.9: 104787880-104787880 on Build GRCh38
rs764456041 Chr.9: 104788068-104788068 on Build GRCh38
rs886063305 Chr.9: 104788015-104788015 on Build GRCh38
rs199604917 Chr.9: 104788035-104788035 on Build GRCh38
rs150283412 Chr.9: 104788006-104788006 on Build GRCh38
rs756549407 Chr.9: 104787961-104787961 on Build GRCh38
rs766619359 Chr.9: 104787931-104787931 on Build GRCh38
rs778400988 Chr.9: 104787972-104787972 on Build GRCh38
rs9282537 Chr.9: 104787941-104787941 on Build GRCh38
rs200788099 Chr.9: 104788041-104788041 on Build GRCh38
rs113373258 Chr.9: 104788022-104788022 on Build GRCh38
rs370225084 Chr.9: 104788091-104788091 on Build GRCh38
rs2020928 Chr.9: 104788078-104788078 on Build GRCh38
rs1043629807 Chr.9: 104787928-104787928 on Build GRCh38
rs746601066 Chr.9: 104787973-104787973 on Build GRCh38
rs369286248 Chr.9: 104787886-104787886 on Build GRCh38
rs565625985 Chr.9: 104787965-104787965 on Build GRCh38
rs760037059 Chr.9: 104788045-104788045 on Build GRCh38
rs576153555 Chr.9: 104788047-104788047 on Build GRCh38

View Full Product Details