Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104741739-104741974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSSL1
Entrez Gene ID
122622
Gene Name
adenylosuccinate synthase like 1
Gene Aliases
-
UniGene
Hs.592327
Gene Chromosome Location
Chr.14: 104724197-104747325 on Build GRCh38
Transcripts
NM_001320424.1 NM_152328.4 NM_199165.2 XM_006720026.3 XM_011536412.1 XM_011536414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595644 c.1009G>A p.V337M Chr.14: 104741934-104741934 on Build GRCh38
5903065 c.989C>T p.P330L Chr.14: 104741914-104741914 on Build GRCh38
5847659 c.937G>T p.V313L Chr.14: 104741862-104741862 on Build GRCh38
1707916 c.1021G>A p.V341M Chr.14: 104741946-104741946 on Build GRCh38
381946 c.960G>T p.V320V Chr.14: 104741885-104741885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767474810 Chr.14: 104741914-104741914 on Build GRCh38
rs750854629 Chr.14: 104741935-104741935 on Build GRCh38
rs780224589 Chr.14: 104741943-104741943 on Build GRCh38
rs45518837 Chr.14: 104741769-104741769 on Build GRCh38
rs142728399 Chr.14: 104741900-104741900 on Build GRCh38
rs1038201766 Chr.14: 104741778-104741778 on Build GRCh38
rs898292702 Chr.14: 104741782-104741782 on Build GRCh38
rs537646950 Chr.14: 104741789-104741789 on Build GRCh38
rs775952636 Chr.14: 104741895-104741895 on Build GRCh38
rs757214349 Chr.14: 104741801-104741801 on Build GRCh38
rs753685530 Chr.14: 104741924-104741924 on Build GRCh38
rs773779986 Chr.14: 104741911-104741911 on Build GRCh38
rs765472066 Chr.14: 104741877-104741877 on Build GRCh38
rs773963466 Chr.14: 104741858-104741859 on Build GRCh38
rs781702955 Chr.14: 104741934-104741934 on Build GRCh38
rs761679828 Chr.14: 104741856-104741856 on Build GRCh38
rs758630677 Chr.14: 104741925-104741925 on Build GRCh38
rs1033687494 Chr.14: 104741897-104741897 on Build GRCh38
rs375399587 Chr.14: 104741831-104741831 on Build GRCh38
rs779066229 Chr.14: 104741810-104741810 on Build GRCh38
rs769476611 Chr.14: 104741837-104741837 on Build GRCh38
rs769266263 Chr.14: 104741918-104741918 on Build GRCh38
rs775055375 Chr.14: 104741841-104741841 on Build GRCh38
rs553526202 Chr.14: 104741942-104741942 on Build GRCh38
rs201545877 Chr.14: 104741811-104741811 on Build GRCh38
rs755917775 Chr.14: 104741875-104741875 on Build GRCh38
rs4983385 Chr.14: 104741792-104741792 on Build GRCh38
rs746286168 Chr.14: 104741835-104741835 on Build GRCh38
rs773069031 Chr.14: 104741916-104741916 on Build GRCh38
rs74089211 Chr.14: 104741928-104741928 on Build GRCh38
rs537860316 Chr.14: 104741848-104741848 on Build GRCh38
rs764479024 Chr.14: 104741926-104741926 on Build GRCh38
rs767443349 Chr.14: 104741858-104741858 on Build GRCh38
rs766041366 Chr.14: 104741921-104741921 on Build GRCh38
rs747472414 Chr.14: 104741818-104741818 on Build GRCh38
rs371777343 Chr.14: 104741917-104741917 on Build GRCh38
rs750224112 Chr.14: 104741861-104741861 on Build GRCh38
rs752824822 Chr.14: 104741882-104741882 on Build GRCh38
rs111588752 Chr.14: 104741846-104741846 on Build GRCh38
rs113582702 Chr.14: 104741932-104741932 on Build GRCh38
rs148388939 Chr.14: 104741933-104741933 on Build GRCh38
rs746858612 Chr.14: 104741913-104741913 on Build GRCh38
rs780914437 Chr.14: 104741804-104741804 on Build GRCh38
rs200862247 Chr.14: 104741816-104741816 on Build GRCh38
rs766015137 Chr.14: 104741816-104741819 on Build GRCh38
rs773873572 Chr.14: 104741853-104741853 on Build GRCh38
rs777945958 Chr.14: 104741886-104741886 on Build GRCh38
rs747094101 Chr.14: 104741887-104741887 on Build GRCh38
rs770096651 Chr.14: 104741892-104741892 on Build GRCh38
rs751537137 Chr.14: 104741799-104741799 on Build GRCh38
rs758718604 Chr.14: 104741883-104741883 on Build GRCh38
rs368655353 Chr.14: 104741851-104741851 on Build GRCh38
rs750195957 Chr.14: 104741809-104741809 on Build GRCh38
rs757835976 Chr.14: 104741888-104741888 on Build GRCh38
rs1054064845 Chr.14: 104741783-104741783 on Build GRCh38
rs542060225 Chr.14: 104741899-104741899 on Build GRCh38
rs574062486 Chr.14: 104741889-104741889 on Build GRCh38
rs771581444 Chr.14: 104741825-104741825 on Build GRCh38
rs1002447823 Chr.14: 104741798-104741798 on Build GRCh38
rs746223893 Chr.14: 104741891-104741891 on Build GRCh38

View Full Product Details