Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 51000626-51000897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM9
Entrez Gene ID
114088
Gene Name
tripartite motif containing 9
Gene Aliases
RNF91, SPRING
UniGene
Hs.654750
Gene Chromosome Location
Chr.14: 50975261-51105173 on Build GRCh38
Transcripts
NM_015163.5 NM_052978.4 XM_005267301.3 XM_005267302.3 XM_006720018.3 XM_011536389.2 XM_017020943.1 XM_017020944.1 XM_017020945.1 XM_017020946.1 XM_017020947.1 XM_017020948.1 XM_017020949.1 XM_017020950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3711500 c.1456C>A p.Q486K Chr.14: 51000691-51000691 on Build GRCh38
3744368 c.1357A>T p.T453S Chr.14: 51000790-51000790 on Build GRCh38
6140437 c.1307-2A>G p.? Chr.14: 51000842-51000842 on Build GRCh38
4051174 c.1365G>A p.T455T Chr.14: 51000770-51000770 on Build GRCh38
347032 c.1402G>T p.A468S Chr.14: 51000733-51000733 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765701217 Chr.14: 51000828-51000828 on Build GRCh38
rs777361081 Chr.14: 51000782-51000782 on Build GRCh38
rs779597108 Chr.14: 51000764-51000764 on Build GRCh38
rs374315245 Chr.14: 51000851-51000851 on Build GRCh38
rs149217157 Chr.14: 51000684-51000684 on Build GRCh38
rs977261155 Chr.14: 51000854-51000854 on Build GRCh38
rs377017804 Chr.14: 51000861-51000861 on Build GRCh38
rs200192609 Chr.14: 51000770-51000770 on Build GRCh38
rs964089850 Chr.14: 51000763-51000763 on Build GRCh38
rs761319285 Chr.14: 51000733-51000733 on Build GRCh38
rs544784389 Chr.14: 51000776-51000776 on Build GRCh38
rs775757427 Chr.14: 51000793-51000793 on Build GRCh38
rs377103808 Chr.14: 51000814-51000814 on Build GRCh38
rs775705971 Chr.14: 51000731-51000731 on Build GRCh38
rs200001803 Chr.14: 51000740-51000740 on Build GRCh38
rs751048253 Chr.14: 51000765-51000765 on Build GRCh38
rs565032018 Chr.14: 51000800-51000800 on Build GRCh38
rs577952155 Chr.14: 51000660-51000660 on Build GRCh38
rs759618741 Chr.14: 51000706-51000706 on Build GRCh38
rs747906315 Chr.14: 51000667-51000667 on Build GRCh38
rs762717966 Chr.14: 51000839-51000839 on Build GRCh38
rs751171171 Chr.14: 51000863-51000863 on Build GRCh38
rs777295762 Chr.14: 51000816-51000816 on Build GRCh38
rs750740333 Chr.14: 51000833-51000833 on Build GRCh38
rs375333472 Chr.14: 51000737-51000737 on Build GRCh38
rs764098743 Chr.14: 51000778-51000780 on Build GRCh38
rs1047251261 Chr.14: 51000676-51000676 on Build GRCh38
rs973334635 Chr.14: 51000781-51000781 on Build GRCh38
rs750095606 Chr.14: 51000752-51000752 on Build GRCh38
rs757983876 Chr.14: 51000754-51000754 on Build GRCh38
rs767636523 Chr.14: 51000730-51000730 on Build GRCh38
rs746166614 Chr.14: 51000790-51000790 on Build GRCh38
rs954491524 Chr.14: 51000662-51000662 on Build GRCh38
rs772167013 Chr.14: 51000791-51000791 on Build GRCh38
rs200506401 Chr.14: 51000697-51000697 on Build GRCh38
rs765465611 Chr.14: 51000741-51000741 on Build GRCh38
rs748027418 Chr.14: 51000775-51000775 on Build GRCh38
rs542776479 Chr.14: 51000734-51000734 on Build GRCh38
rs762163337 Chr.14: 51000827-51000827 on Build GRCh38
rs887395439 Chr.14: 51000696-51000696 on Build GRCh38
rs372743437 Chr.14: 51000663-51000663 on Build GRCh38

View Full Product Details