Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39265768-39266027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1 NM_001337.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3593882 c.696C>T p.F232F Chr.3: 39265910-39265910 on Build GRCh38
3660543 c.651G>A p.T217T Chr.3: 39265859-39265859 on Build GRCh38
3593878 c.785T>A p.I262N Chr.3: 39265821-39265821 on Build GRCh38
2987589 c.553G>A p.E185K Chr.3: 39265957-39265957 on Build GRCh38
6026793 c.582A>G p.A194A Chr.3: 39266024-39266024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868320190 Chr.3: 39265981-39265981 on Build GRCh38
rs752215576 Chr.3: 39266002-39266002 on Build GRCh38
rs780606575 Chr.3: 39265898-39265898 on Build GRCh38
rs149456270 Chr.3: 39266003-39266003 on Build GRCh38
rs766834140 Chr.3: 39265969-39265969 on Build GRCh38
rs376609543 Chr.3: 39265829-39265829 on Build GRCh38
rs765623786 Chr.3: 39265841-39265841 on Build GRCh38
rs200607201 Chr.3: 39265939-39265939 on Build GRCh38
rs752076939 Chr.3: 39265970-39265970 on Build GRCh38
rs775640906 Chr.3: 39265795-39265795 on Build GRCh38
rs928981544 Chr.3: 39265851-39265851 on Build GRCh38
rs770929576 Chr.3: 39265933-39265933 on Build GRCh38
rs376411124 Chr.3: 39265934-39265934 on Build GRCh38
rs773298308 Chr.3: 39265946-39265946 on Build GRCh38
rs4986872 Chr.3: 39265853-39265853 on Build GRCh38
rs760009245 Chr.3: 39265979-39265979 on Build GRCh38
rs183530704 Chr.3: 39265840-39265840 on Build GRCh38
rs956057885 Chr.3: 39265912-39265912 on Build GRCh38
rs751023067 Chr.3: 39265870-39265870 on Build GRCh38
rs865941167 Chr.3: 39265947-39265947 on Build GRCh38
rs201878012 Chr.3: 39265992-39265992 on Build GRCh38
rs868111811 Chr.3: 39265864-39265864 on Build GRCh38
rs760818722 Chr.3: 39265803-39265803 on Build GRCh38
rs749013292 Chr.3: 39265842-39265842 on Build GRCh38
rs928825762 Chr.3: 39265954-39265954 on Build GRCh38
rs776940684 Chr.3: 39265830-39265830 on Build GRCh38
rs897723430 Chr.3: 39265847-39265847 on Build GRCh38
rs754549648 Chr.3: 39265889-39265889 on Build GRCh38
rs369705104 Chr.3: 39265916-39265916 on Build GRCh38
rs777578095 Chr.3: 39266007-39266007 on Build GRCh38
rs752371213 Chr.3: 39265910-39265910 on Build GRCh38
rs747017950 Chr.3: 39265937-39265937 on Build GRCh38
rs768536284 Chr.3: 39265902-39265902 on Build GRCh38
rs924631311 Chr.3: 39265883-39265883 on Build GRCh38
rs749096233 Chr.3: 39265931-39265931 on Build GRCh38
rs756353554 Chr.3: 39265835-39265835 on Build GRCh38
rs773508118 Chr.3: 39265945-39265945 on Build GRCh38
rs374664955 Chr.3: 39265860-39265860 on Build GRCh38
rs376455816 Chr.3: 39265863-39265863 on Build GRCh38
rs768761417 Chr.3: 39265938-39265938 on Build GRCh38
rs373292400 Chr.3: 39265918-39265918 on Build GRCh38
rs956955089 Chr.3: 39265948-39265948 on Build GRCh38

View Full Product Details