Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132079923-132080178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs1017374361 Chr.X: 132079980-132079980 on Build GRCh38
rs753705251 Chr.X: 132079969-132079969 on Build GRCh38
rs983334670 Chr.X: 132080105-132080105 on Build GRCh38
rs762011171 Chr.X: 132080110-132080110 on Build GRCh38
rs137852208 Chr.X: 132080053-132080053 on Build GRCh38
rs994669070 Chr.X: 132080066-132080066 on Build GRCh38
rs752370008 Chr.X: 132079994-132079994 on Build GRCh38
rs781757793 Chr.X: 132080058-132080058 on Build GRCh38
rs778646607 Chr.X: 132079984-132079984 on Build GRCh38
rs777152565 Chr.X: 132080016-132080016 on Build GRCh38
rs773298405 Chr.X: 132080064-132080064 on Build GRCh38
rs966404639 Chr.X: 132079986-132079986 on Build GRCh38
rs758009021 Chr.X: 132080000-132080000 on Build GRCh38
rs149159499 Chr.X: 132080069-132080069 on Build GRCh38
rs571080820 Chr.X: 132080112-132080112 on Build GRCh38
rs761046859 Chr.X: 132080073-132080075 on Build GRCh38
rs774307938 Chr.X: 132080097-132080097 on Build GRCh38
rs767487972 Chr.X: 132080147-132080147 on Build GRCh38
rs370437110 Chr.X: 132080134-132080134 on Build GRCh38
rs950634229 Chr.X: 132080071-132080071 on Build GRCh38
rs754832399 Chr.X: 132079974-132079974 on Build GRCh38
rs145630934 Chr.X: 132080073-132080073 on Build GRCh38
rs111377071 Chr.X: 132080111-132080111 on Build GRCh38
rs769410120 Chr.X: 132080052-132080052 on Build GRCh38
rs770328478 Chr.X: 132080061-132080061 on Build GRCh38

View Full Product Details