Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33203684-33203917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC39A7
Entrez Gene ID
7922
Gene Name
solute carrier family 39 member 7
Gene Aliases
D6S115E, D6S2244E, H2-KE4, HKE4, KE4, RING5, ZIP7
UniGene
Hs.631995
Gene Chromosome Location
Chr.6: 33200826-33204437 on Build GRCh38
Transcripts
NM_001077516.1 NM_001288777.1 NM_006979.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4727355 c.1368delG p.G458fs*4 Chr.6: 33203771-33203771 on Build GRCh38
7094750 c.1312G>A p.E438K Chr.6: 33203715-33203715 on Build GRCh38
6448447 c.1288G>A p.V430I Chr.6: 33203691-33203691 on Build GRCh38
4727354 c.1291G>T p.A431S Chr.6: 33203694-33203694 on Build GRCh38
1329764 c.1306T>G p.L436V Chr.6: 33203709-33203709 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927259802 Chr.6: 33203879-33203879 on Build GRCh38
rs369067386 Chr.6: 33203714-33203714 on Build GRCh38
rs372748339 Chr.6: 33203773-33203773 on Build GRCh38
rs747721150 Chr.6: 33203857-33203857 on Build GRCh38
rs778523225 Chr.6: 33203744-33203744 on Build GRCh38
rs764190480 Chr.6: 33203782-33203782 on Build GRCh38
rs369573351 Chr.6: 33203817-33203817 on Build GRCh38
rs373995549 Chr.6: 33203854-33203854 on Build GRCh38
rs761933592 Chr.6: 33203798-33203798 on Build GRCh38
rs774595475 Chr.6: 33203781-33203783 on Build GRCh38
rs755206565 Chr.6: 33203871-33203871 on Build GRCh38
rs771668497 Chr.6: 33203764-33203764 on Build GRCh38
rs957742635 Chr.6: 33203868-33203868 on Build GRCh38
rs548487329 Chr.6: 33203803-33203803 on Build GRCh38
rs780518091 Chr.6: 33203845-33203845 on Build GRCh38
rs372930762 Chr.6: 33203861-33203861 on Build GRCh38
rs377655534 Chr.6: 33203855-33203855 on Build GRCh38
rs374131623 Chr.6: 33203872-33203872 on Build GRCh38
rs747627669 Chr.6: 33203755-33203755 on Build GRCh38
rs751680391 Chr.6: 33203784-33203784 on Build GRCh38
rs369084525 Chr.6: 33203859-33203859 on Build GRCh38
rs907750113 Chr.6: 33203830-33203830 on Build GRCh38
rs756494407 Chr.6: 33203731-33203731 on Build GRCh38
rs530289508 Chr.6: 33203739-33203739 on Build GRCh38
rs767698096 Chr.6: 33203799-33203799 on Build GRCh38
rs923056227 Chr.6: 33203760-33203760 on Build GRCh38
rs780428908 Chr.6: 33203732-33203732 on Build GRCh38
rs772908349 Chr.6: 33203772-33203772 on Build GRCh38
rs375996215 Chr.6: 33203776-33203776 on Build GRCh38
rs749753724 Chr.6: 33203735-33203735 on Build GRCh38
rs200525744 Chr.6: 33203725-33203725 on Build GRCh38

View Full Product Details