Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125855043-125855274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
UniGene
Hs.171426
Gene Chromosome Location
Chr.6: 125780948-125932030 on Build GRCh38
Transcripts
NM_001122842.2 NM_001199619.1 NM_001199620.1 NM_001199621.1 NM_181782.4 XM_005266822.4 XM_006715340.3 XM_011535455.2 XM_017010269.1 XM_017010270.1 XM_017010271.1 XM_017010272.1 XM_017010273.1 XM_017010274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325926 c.159C>A p.D53E Chr.6: 125855128-125855128 on Build GRCh38
1440253 c.201_202insA p.R71fs*4 Chr.6: 125855170-125855171 on Build GRCh38
739333 c.238A>G p.R80G Chr.6: 125855207-125855207 on Build GRCh38
6235946 c.210G>A p.K70K Chr.6: 125855179-125855179 on Build GRCh38
3393794 c.242G>A p.R81H Chr.6: 125855211-125855211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781288780 Chr.6: 125855069-125855069 on Build GRCh38
rs747877767 Chr.6: 125855071-125855071 on Build GRCh38
rs574387361 Chr.6: 125855105-125855105 on Build GRCh38
rs749319230 Chr.6: 125855171-125855171 on Build GRCh38
rs779767564 Chr.6: 125855210-125855210 on Build GRCh38
rs766964758 Chr.6: 125855170-125855170 on Build GRCh38
rs771407165 Chr.6: 125855237-125855237 on Build GRCh38
rs148461613 Chr.6: 125855143-125855143 on Build GRCh38
rs78815147 Chr.6: 125855178-125855178 on Build GRCh38
rs755904508 Chr.6: 125855072-125855072 on Build GRCh38
rs766825580 Chr.6: 125855147-125855147 on Build GRCh38
rs1025245959 Chr.6: 125855073-125855073 on Build GRCh38
rs770852055 Chr.6: 125855187-125855187 on Build GRCh38
rs142658591 Chr.6: 125855155-125855155 on Build GRCh38
rs749213685 Chr.6: 125855078-125855078 on Build GRCh38
rs150585521 Chr.6: 125855204-125855204 on Build GRCh38
rs945056668 Chr.6: 125855190-125855190 on Build GRCh38
rs971881607 Chr.6: 125855175-125855175 on Build GRCh38
rs757180215 Chr.6: 125855182-125855182 on Build GRCh38
rs773759641 Chr.6: 125855137-125855137 on Build GRCh38
rs980968621 Chr.6: 125855110-125855110 on Build GRCh38
rs369455623 Chr.6: 125855227-125855227 on Build GRCh38
rs773883686 Chr.6: 125855084-125855084 on Build GRCh38
rs756388962 Chr.6: 125855158-125855158 on Build GRCh38
rs746636348 Chr.6: 125855211-125855211 on Build GRCh38
rs763428656 Chr.6: 125855234-125855234 on Build GRCh38
rs777525135 Chr.6: 125855163-125855163 on Build GRCh38
rs762320855 Chr.6: 125855248-125855249 on Build GRCh38
rs771705852 Chr.6: 125855099-125855099 on Build GRCh38
rs529214502 Chr.6: 125855100-125855100 on Build GRCh38
rs752909312 Chr.6: 125855157-125855157 on Build GRCh38
rs769322345 Chr.6: 125855233-125855234 on Build GRCh38
rs745402203 Chr.6: 125855086-125855086 on Build GRCh38
rs989304683 Chr.6: 125855160-125855160 on Build GRCh38
rs745327382 Chr.6: 125855200-125855200 on Build GRCh38
rs375340145 Chr.6: 125855090-125855090 on Build GRCh38
rs751510610 Chr.6: 125855075-125855075 on Build GRCh38
rs987770322 Chr.6: 125855202-125855202 on Build GRCh38
rs770909278 Chr.6: 125855082-125855082 on Build GRCh38
rs1033837864 Chr.6: 125855133-125855133 on Build GRCh38
rs767709968 Chr.6: 125855156-125855156 on Build GRCh38
rs147393001 Chr.6: 125855124-125855124 on Build GRCh38
rs765682345 Chr.6: 125855136-125855136 on Build GRCh38
rs763522495 Chr.6: 125855138-125855138 on Build GRCh38
rs768462504 Chr.6: 125855214-125855214 on Build GRCh38
rs774722148 Chr.6: 125855250-125855250 on Build GRCh38

View Full Product Details