Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12156745-12157018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYN2
Entrez Gene ID
6854
Gene Name
synapsin II
Gene Aliases
SYNII
UniGene
Hs.445503
Gene Chromosome Location
Chr.3: 12004360-12192032 on Build GRCh38
Transcripts
NM_003178.5 NM_133625.4 XM_006713311.3 XM_006713312.3 XM_006713313.2 NM_003256.3

Gene Symbol
TIMP4
Entrez Gene ID
7079
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
UniGene
Hs.591665
Gene Chromosome Location
Chr.3: 12153068-12159351 on Build GRCh38
Transcripts
NM_003178.5 NM_133625.4 XM_006713311.3 XM_006713312.3 XM_006713313.2 NM_003256.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4935644 c.306T>C p.C102C Chr.3: 12156866-12156866 on Build GRCh38
6276499 c.337C>A p.Q113K Chr.3: 12156835-12156835 on Build GRCh38
5814623 c.328A>T p.S110C Chr.3: 12156844-12156844 on Build GRCh38
343014 c.240G>C p.M80I Chr.3: 12156932-12156932 on Build GRCh38
1744552 c.279C>G p.I93M Chr.3: 12156893-12156893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745773299 Chr.3: 12156862-12156862 on Build GRCh38
rs757919305 Chr.3: 12156980-12156980 on Build GRCh38
rs560908062 Chr.3: 12156795-12156795 on Build GRCh38
rs771959850 Chr.3: 12156866-12156866 on Build GRCh38
rs769898540 Chr.3: 12156897-12156897 on Build GRCh38
rs1051736484 Chr.3: 12156908-12156908 on Build GRCh38
rs767828618 Chr.3: 12156950-12156950 on Build GRCh38
rs748770830 Chr.3: 12156778-12156778 on Build GRCh38
rs370191065 Chr.3: 12156789-12156789 on Build GRCh38
rs551555922 Chr.3: 12156865-12156865 on Build GRCh38
rs529579724 Chr.3: 12156812-12156812 on Build GRCh38
rs759971872 Chr.3: 12156939-12156939 on Build GRCh38
rs541277708 Chr.3: 12156837-12156837 on Build GRCh38
rs776102792 Chr.3: 12156949-12156949 on Build GRCh38
rs192507347 Chr.3: 12156847-12156847 on Build GRCh38
rs778657739 Chr.3: 12156964-12156964 on Build GRCh38
rs759512231 Chr.3: 12156791-12156791 on Build GRCh38
rs777309788 Chr.3: 12156771-12156771 on Build GRCh38
rs140467141 Chr.3: 12156925-12156925 on Build GRCh38
rs746749753 Chr.3: 12156885-12156885 on Build GRCh38
rs751920911 Chr.3: 12156841-12156841 on Build GRCh38
rs759026154 Chr.3: 12156836-12156836 on Build GRCh38
rs750932059 Chr.3: 12156832-12156832 on Build GRCh38
rs531019174 Chr.3: 12156938-12156938 on Build GRCh38
rs772714164 Chr.3: 12156794-12156794 on Build GRCh38
rs936069341 Chr.3: 12156948-12156948 on Build GRCh38
rs201423861 Chr.3: 12156888-12156888 on Build GRCh38
rs188059959 Chr.3: 12156860-12156860 on Build GRCh38
rs942194324 Chr.3: 12156843-12156843 on Build GRCh38
rs1001795055 Chr.3: 12156961-12156961 on Build GRCh38
rs777413604 Chr.3: 12156846-12156846 on Build GRCh38
rs370491983 Chr.3: 12156911-12156911 on Build GRCh38
rs373683527 Chr.3: 12156887-12156887 on Build GRCh38
rs756779101 Chr.3: 12156849-12156849 on Build GRCh38
rs10433537 Chr.3: 12156985-12156985 on Build GRCh38
rs753506025 Chr.3: 12156955-12156955 on Build GRCh38
rs745849653 Chr.3: 12156799-12156799 on Build GRCh38
rs779990008 Chr.3: 12156882-12156882 on Build GRCh38
rs749887551 Chr.3: 12156965-12156965 on Build GRCh38
rs1048359264 Chr.3: 12156809-12156809 on Build GRCh38
rs533386083 Chr.3: 12156854-12156854 on Build GRCh38
rs765305630 Chr.3: 12156830-12156830 on Build GRCh38
rs760496338 Chr.3: 12156816-12156816 on Build GRCh38
rs771831162 Chr.3: 12156801-12156801 on Build GRCh38
rs779900052 Chr.3: 12156981-12156981 on Build GRCh38
rs776485206 Chr.3: 12156776-12156776 on Build GRCh38
rs755404721 Chr.3: 12156845-12156845 on Build GRCh38
rs756898684 Chr.3: 12156958-12156958 on Build GRCh38
rs766932798 Chr.3: 12156921-12156921 on Build GRCh38
rs761886594 Chr.3: 12156829-12156829 on Build GRCh38
rs1039262990 Chr.3: 12156877-12156877 on Build GRCh38
rs776732816 Chr.3: 12156828-12156828 on Build GRCh38
rs972976983 Chr.3: 12156794-12156794 on Build GRCh38
rs764253609 Chr.3: 12156820-12156820 on Build GRCh38
rs199719951 Chr.3: 12156899-12156899 on Build GRCh38

View Full Product Details