Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27150126-27150385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCF23
Entrez Gene ID
150921
Gene Name
transcription factor 23
Gene Aliases
OUT, TCF-23, bHLHa24
UniGene
Hs.591583
Gene Chromosome Location
Chr.2: 27148895-27153577 on Build GRCh38
Transcripts
NM_175769.2 XM_005264159.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1668789 c.361G>A p.A121T Chr.2: 27150261-27150261 on Build GRCh38
4732681 c.437G>A p.R146H Chr.2: 27150337-27150337 on Build GRCh38
1636946 c.277C>T p.R93C Chr.2: 27150177-27150177 on Build GRCh38
4732680 c.348C>T p.D116D Chr.2: 27150248-27150248 on Build GRCh38
4768423 c.384C>A p.L128L Chr.2: 27150284-27150284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757041920 Chr.2: 27150261-27150261 on Build GRCh38
rs746191605 Chr.2: 27150354-27150354 on Build GRCh38
rs776758564 Chr.2: 27150209-27150209 on Build GRCh38
rs373445415 Chr.2: 27150366-27150366 on Build GRCh38
rs146000022 Chr.2: 27150337-27150337 on Build GRCh38
rs148091174 Chr.2: 27150298-27150298 on Build GRCh38
rs773919595 Chr.2: 27150234-27150234 on Build GRCh38
rs1015479249 Chr.2: 27150257-27150257 on Build GRCh38
rs138997726 Chr.2: 27150174-27150174 on Build GRCh38
rs759113306 Chr.2: 27150297-27150297 on Build GRCh38
rs761798617 Chr.2: 27150321-27150321 on Build GRCh38
rs906023521 Chr.2: 27150328-27150328 on Build GRCh38
rs1002946243 Chr.2: 27150221-27150221 on Build GRCh38
rs755225473 Chr.2: 27150267-27150267 on Build GRCh38
rs369275879 Chr.2: 27150150-27150150 on Build GRCh38
rs914799086 Chr.2: 27150320-27150320 on Build GRCh38
rs200237600 Chr.2: 27150224-27150224 on Build GRCh38
rs762601345 Chr.2: 27150147-27150147 on Build GRCh38
rs771652114 Chr.2: 27150359-27150359 on Build GRCh38
rs755171054 Chr.2: 27150335-27150335 on Build GRCh38
rs748722467 Chr.2: 27150215-27150215 on Build GRCh38
rs1050740688 Chr.2: 27150159-27150159 on Build GRCh38
rs777912117 Chr.2: 27150342-27150342 on Build GRCh38
rs758714524 Chr.2: 27150173-27150173 on Build GRCh38
rs1053368647 Chr.2: 27150177-27150177 on Build GRCh38
rs771398931 Chr.2: 27150345-27150345 on Build GRCh38
rs905971179 Chr.2: 27150219-27150219 on Build GRCh38
rs953823726 Chr.2: 27150275-27150275 on Build GRCh38
rs201667688 Chr.2: 27150178-27150178 on Build GRCh38
rs1011669756 Chr.2: 27150145-27150145 on Build GRCh38
rs370405355 Chr.2: 27150260-27150260 on Build GRCh38
rs75689774 Chr.2: 27150286-27150286 on Build GRCh38
rs756231299 Chr.2: 27150339-27150339 on Build GRCh38
rs150714023 Chr.2: 27150163-27150163 on Build GRCh38
rs778217239 Chr.2: 27150292-27150292 on Build GRCh38
rs146034019 Chr.2: 27150149-27150149 on Build GRCh38
rs144811550 Chr.2: 27150199-27150199 on Build GRCh38
rs753875573 Chr.2: 27150319-27150319 on Build GRCh38
rs184207079 Chr.2: 27150172-27150172 on Build GRCh38
rs377184068 Chr.2: 27150164-27150164 on Build GRCh38
rs755486123 Chr.2: 27150148-27150148 on Build GRCh38
rs139709058 Chr.2: 27150185-27150185 on Build GRCh38
rs376284002 Chr.2: 27150248-27150248 on Build GRCh38
rs759209676 Chr.2: 27150236-27150236 on Build GRCh38
rs770310534 Chr.2: 27150290-27150290 on Build GRCh38
rs1044613600 Chr.2: 27150326-27150326 on Build GRCh38
rs373573959 Chr.2: 27150296-27150296 on Build GRCh38
rs763666809 Chr.2: 27150313-27150313 on Build GRCh38
rs778747753 Chr.2: 27150263-27150263 on Build GRCh38
rs748170605 Chr.2: 27150288-27150288 on Build GRCh38
rs149001386 Chr.2: 27150216-27150216 on Build GRCh38
rs766999123 Chr.2: 27150239-27150239 on Build GRCh38
rs533332704 Chr.2: 27150325-27150325 on Build GRCh38
rs550752748 Chr.2: 27150293-27150293 on Build GRCh38
rs138845716 Chr.2: 27150303-27150303 on Build GRCh38
rs763257227 Chr.2: 27150220-27150220 on Build GRCh38
rs752807433 Chr.2: 27150338-27150338 on Build GRCh38
rs376939643 Chr.2: 27150249-27150249 on Build GRCh38
rs777811766 Chr.2: 27150341-27150341 on Build GRCh38
rs773563171 Chr.2: 27150146-27150146 on Build GRCh38
rs574334375 Chr.2: 27150346-27150346 on Build GRCh38
rs767027596 Chr.2: 27150157-27150157 on Build GRCh38
rs550968155 Chr.2: 27150312-27150312 on Build GRCh38
rs750168811 Chr.2: 27150333-27150333 on Build GRCh38
rs199691868 Chr.2: 27150264-27150264 on Build GRCh38
rs752736196 Chr.2: 27150162-27150162 on Build GRCh38
rs529401580 Chr.2: 27150277-27150277 on Build GRCh38

View Full Product Details