Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2519485-2519730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0C
Entrez Gene ID
527
Gene Name
ATPase H+ transporting V0 subunit c
Gene Aliases
ATP6C, ATP6L, ATPL, VATL, VPPC, Vma3
UniGene
Hs.389107
Gene Chromosome Location
Chr.16: 2513726-2520223 on Build GRCh38
Transcripts
NM_001198569.1 NM_001694.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5701492 c.316_317insGCCGGCTTT p.A106>GRLS Chr.16: 2519593-2519594 on Build GRCh38
3108470 c.282C>T p.G94G Chr.16: 2519559-2519559 on Build GRCh38
5483317 c.424G>A p.G142S Chr.16: 2519701-2519701 on Build GRCh38
6880315 c.431A>G p.Y144C Chr.16: 2519708-2519708 on Build GRCh38
1678782 c.358G>T p.G120C Chr.16: 2519635-2519635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764908287 Chr.16: 2519566-2519566 on Build GRCh38
rs781117999 Chr.16: 2519589-2519589 on Build GRCh38
rs751318726 Chr.16: 2519583-2519583 on Build GRCh38
rs201568963 Chr.16: 2519612-2519612 on Build GRCh38
rs11546344 Chr.16: 2519710-2519710 on Build GRCh38
rs752604908 Chr.16: 2519709-2519709 on Build GRCh38
rs376971724 Chr.16: 2519505-2519505 on Build GRCh38
rs201431472 Chr.16: 2519580-2519580 on Build GRCh38
rs1044901385 Chr.16: 2519586-2519586 on Build GRCh38
rs781013495 Chr.16: 2519522-2519522 on Build GRCh38
rs749478981 Chr.16: 2519616-2519616 on Build GRCh38
rs556217954 Chr.16: 2519559-2519559 on Build GRCh38
rs200837309 Chr.16: 2519694-2519694 on Build GRCh38
rs769866122 Chr.16: 2519526-2519526 on Build GRCh38
rs761323538 Chr.16: 2519661-2519661 on Build GRCh38
rs777875089 Chr.16: 2519613-2519613 on Build GRCh38
rs758265268 Chr.16: 2519518-2519518 on Build GRCh38
rs761344058 Chr.16: 2519562-2519562 on Build GRCh38
rs149144429 Chr.16: 2519622-2519622 on Build GRCh38
rs370823612 Chr.16: 2519691-2519691 on Build GRCh38
rs773415809 Chr.16: 2519527-2519527 on Build GRCh38
rs753352515 Chr.16: 2519503-2519503 on Build GRCh38
rs772332489 Chr.16: 2519631-2519631 on Build GRCh38
rs754740949 Chr.16: 2519584-2519584 on Build GRCh38
rs748200806 Chr.16: 2519523-2519523 on Build GRCh38
rs746899524 Chr.16: 2519520-2519520 on Build GRCh38
rs750086896 Chr.16: 2519571-2519571 on Build GRCh38
rs756170092 Chr.16: 2519601-2519601 on Build GRCh38
rs771258996 Chr.16: 2519533-2519533 on Build GRCh38
rs752697033 Chr.16: 2519595-2519595 on Build GRCh38
rs143553028 Chr.16: 2519673-2519673 on Build GRCh38
rs751174146 Chr.16: 2519700-2519700 on Build GRCh38
rs11546342 Chr.16: 2519617-2519617 on Build GRCh38
rs536381963 Chr.16: 2519534-2519534 on Build GRCh38
rs775877366 Chr.16: 2519561-2519561 on Build GRCh38
rs776063145 Chr.16: 2519640-2519640 on Build GRCh38
rs146819964 Chr.16: 2519703-2519703 on Build GRCh38
rs750107413 Chr.16: 2519514-2519514 on Build GRCh38
rs757038421 Chr.16: 2519585-2519585 on Build GRCh38
rs948026839 Chr.16: 2519664-2519664 on Build GRCh38
rs765996446 Chr.16: 2519697-2519697 on Build GRCh38
rs749567124 Chr.16: 2519532-2519532 on Build GRCh38
rs779029141 Chr.16: 2519623-2519623 on Build GRCh38
rs764928515 Chr.16: 2519507-2519507 on Build GRCh38
rs780055661 Chr.16: 2519519-2519519 on Build GRCh38
rs759853764 Chr.16: 2519553-2519553 on Build GRCh38
rs746189093 Chr.16: 2519628-2519628 on Build GRCh38

View Full Product Details