Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58611875-58612116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM10
Entrez Gene ID
102
Gene Name
ADAM metallopeptidase domain 10
Gene Aliases
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz
UniGene
Hs.172028
Gene Chromosome Location
Chr.15: 58595204-58749978 on Build GRCh38
Transcripts
NM_001110.3 NM_001320570.1 XM_017021861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400045 c.1512-4C>G p.? Chr.15: 58611995-58611995 on Build GRCh38
5000681 c.1628G>T p.C543F Chr.15: 58611875-58611875 on Build GRCh38
470876 c.1625T>C p.L542P Chr.15: 58611878-58611878 on Build GRCh38
4896926 c.1612G>A p.G538S Chr.15: 58611891-58611891 on Build GRCh38
1301260 c.1579G>A p.D527N Chr.15: 58611924-58611924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774843170 Chr.15: 58611898-58611898 on Build GRCh38
rs771542514 Chr.15: 58612043-58612043 on Build GRCh38
rs759309487 Chr.15: 58611902-58611902 on Build GRCh38
rs756019865 Chr.15: 58611904-58611904 on Build GRCh38
rs200902548 Chr.15: 58611977-58611977 on Build GRCh38
rs760123663 Chr.15: 58611952-58611952 on Build GRCh38
rs750200666 Chr.15: 58611995-58611995 on Build GRCh38
rs200794475 Chr.15: 58611926-58611926 on Build GRCh38
rs201621495 Chr.15: 58611938-58611938 on Build GRCh38
rs371445663 Chr.15: 58611922-58611922 on Build GRCh38
rs531204671 Chr.15: 58612079-58612079 on Build GRCh38
rs367915349 Chr.15: 58611967-58611967 on Build GRCh38
rs749866094 Chr.15: 58612040-58612040 on Build GRCh38
rs983276222 Chr.15: 58611964-58611964 on Build GRCh38
rs756163425 Chr.15: 58612037-58612037 on Build GRCh38
rs201426430 Chr.15: 58612021-58612021 on Build GRCh38
rs568909884 Chr.15: 58612065-58612065 on Build GRCh38
rs199959350 Chr.15: 58611957-58611957 on Build GRCh38
rs911853783 Chr.15: 58612032-58612032 on Build GRCh38
rs8039791 Chr.15: 58612038-58612038 on Build GRCh38
rs202221661 Chr.15: 58612033-58612033 on Build GRCh38
rs202147172 Chr.15: 58611996-58611996 on Build GRCh38
rs765078645 Chr.15: 58611985-58611985 on Build GRCh38
rs483352916 Chr.15: 58611932-58611932 on Build GRCh38
rs755268496 Chr.15: 58612000-58612000 on Build GRCh38
rs767042249 Chr.15: 58611906-58611906 on Build GRCh38
rs200316504 Chr.15: 58612009-58612009 on Build GRCh38
rs781300827 Chr.15: 58612024-58612024 on Build GRCh38

View Full Product Details