Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37104339-37104594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1A1L
Entrez Gene ID
122011
Gene Name
casein kinase 1 alpha 1 like
Gene Aliases
CK1
UniGene
Hs.512897
Gene Chromosome Location
Chr.13: 37103260-37105664 on Build GRCh38
Transcripts
NM_145203.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636711 c.799C>T p.R267C Chr.13: 37104458-37104458 on Build GRCh38
432365 c.874T>C p.Y292H Chr.13: 37104383-37104383 on Build GRCh38
317200 c.889G>T p.D297Y Chr.13: 37104368-37104368 on Build GRCh38
333917 c.780G>C p.L260F Chr.13: 37104477-37104477 on Build GRCh38
3384831 c.893G>A p.W298* Chr.13: 37104364-37104364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769556012 Chr.13: 37104382-37104382 on Build GRCh38
rs776931065 Chr.13: 37104506-37104506 on Build GRCh38
rs748719787 Chr.13: 37104567-37104567 on Build GRCh38
rs766718454 Chr.13: 37104406-37104406 on Build GRCh38
rs769863283 Chr.13: 37104366-37104366 on Build GRCh38
rs780536500 Chr.13: 37104483-37104484 on Build GRCh38
rs553829073 Chr.13: 37104414-37104414 on Build GRCh38
rs761083671 Chr.13: 37104400-37104400 on Build GRCh38
rs564273735 Chr.13: 37104545-37104545 on Build GRCh38
rs759977373 Chr.13: 37104412-37104412 on Build GRCh38
rs148557143 Chr.13: 37104481-37104481 on Build GRCh38
rs546173732 Chr.13: 37104457-37104457 on Build GRCh38
rs535577422 Chr.13: 37104364-37104364 on Build GRCh38
rs1045864859 Chr.13: 37104396-37104396 on Build GRCh38
rs56252523 Chr.13: 37104567-37104567 on Build GRCh38
rs747425670 Chr.13: 37104361-37104361 on Build GRCh38
rs986825448 Chr.13: 37104560-37104560 on Build GRCh38
rs548321174 Chr.13: 37104553-37104553 on Build GRCh38
rs757887483 Chr.13: 37104433-37104433 on Build GRCh38
rs771196850 Chr.13: 37104503-37104503 on Build GRCh38
rs752163482 Chr.13: 37104413-37104413 on Build GRCh38
rs1011914700 Chr.13: 37104432-37104432 on Build GRCh38
rs773638205 Chr.13: 37104434-37104434 on Build GRCh38
rs878993666 Chr.13: 37104445-37104445 on Build GRCh38
rs878923862 Chr.13: 37104459-37104459 on Build GRCh38
rs759925987 Chr.13: 37104513-37104513 on Build GRCh38
rs751229438 Chr.13: 37104449-37104449 on Build GRCh38
rs752037345 Chr.13: 37104525-37104525 on Build GRCh38
rs55895045 Chr.13: 37104488-37104488 on Build GRCh38
rs780176930 Chr.13: 37104475-37104475 on Build GRCh38
rs763581579 Chr.13: 37104561-37104561 on Build GRCh38
rs893011528 Chr.13: 37104511-37104511 on Build GRCh38
rs368855406 Chr.13: 37104529-37104529 on Build GRCh38
rs756071727 Chr.13: 37104458-37104458 on Build GRCh38
rs762010655 Chr.13: 37104383-37104383 on Build GRCh38
rs755499267 Chr.13: 37104525-37104526 on Build GRCh38
rs1020217324 Chr.13: 37104549-37104549 on Build GRCh38
rs748233566 Chr.13: 37104476-37104476 on Build GRCh38
rs142999830 Chr.13: 37104546-37104546 on Build GRCh38
rs776982240 Chr.13: 37104407-37104407 on Build GRCh38
rs765702339 Chr.13: 37104514-37104514 on Build GRCh38
rs758177403 Chr.13: 37104462-37104462 on Build GRCh38
rs1010136781 Chr.13: 37104528-37104528 on Build GRCh38
rs769712615 Chr.13: 37104566-37104567 on Build GRCh38
rs546656835 Chr.13: 37104419-37104421 on Build GRCh38
rs144553773 Chr.13: 37104444-37104444 on Build GRCh38
rs773476964 Chr.13: 37104484-37104484 on Build GRCh38
rs757073923 Chr.13: 37104452-37104452 on Build GRCh38
rs776027675 Chr.13: 37104550-37104550 on Build GRCh38
rs995862874 Chr.13: 37104479-37104479 on Build GRCh38
rs772302382 Chr.13: 37104387-37104387 on Build GRCh38
rs954465264 Chr.13: 37104512-37104512 on Build GRCh38
rs781495035 Chr.13: 37104536-37104538 on Build GRCh38
rs143083898 Chr.13: 37104388-37104388 on Build GRCh38
rs781072113 Chr.13: 37104456-37104456 on Build GRCh38

View Full Product Details