Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21765807-21766064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ8
Entrez Gene ID
3764
Gene Name
potassium voltage-gated channel subfamily J member 8
Gene Aliases
KIR6.1, uKATP-1
UniGene
Hs.102308
Gene Chromosome Location
Chr.12: 21764955-21775593 on Build GRCh38
Transcripts
NM_004982.3 XM_005253358.4 XM_017019283.1 XM_017019284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224423 c.960G>A p.W320* Chr.12: 21766038-21766038 on Build GRCh38
1177064 c.1136C>A p.S379Y Chr.12: 21765862-21765862 on Build GRCh38
7154638 c.994G>A p.E332K Chr.12: 21766004-21766004 on Build GRCh38
4040827 c.1020C>A p.S340S Chr.12: 21765978-21765978 on Build GRCh38
3980958 c.1182G>A p.R394R Chr.12: 21765816-21765816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754575556 Chr.12: 21765931-21765931 on Build GRCh38
rs757713514 Chr.12: 21765894-21765894 on Build GRCh38
rs147316959 Chr.12: 21765962-21765962 on Build GRCh38
rs974908995 Chr.12: 21765876-21765876 on Build GRCh38
rs34811413 Chr.12: 21765997-21765997 on Build GRCh38
rs778531267 Chr.12: 21765932-21765932 on Build GRCh38
rs746156853 Chr.12: 21766001-21766001 on Build GRCh38
rs147926739 Chr.12: 21766041-21766041 on Build GRCh38
rs775816433 Chr.12: 21766013-21766013 on Build GRCh38
rs770763433 Chr.12: 21765966-21765966 on Build GRCh38
rs752063865 Chr.12: 21765853-21765853 on Build GRCh38
rs764215606 Chr.12: 21766031-21766031 on Build GRCh38
rs930402902 Chr.12: 21765972-21765972 on Build GRCh38
rs867403064 Chr.12: 21765895-21765895 on Build GRCh38
rs769878572 Chr.12: 21766011-21766011 on Build GRCh38
rs764730358 Chr.12: 21765850-21765850 on Build GRCh38
rs777705394 Chr.12: 21765951-21765951 on Build GRCh38
rs374507305 Chr.12: 21765924-21765924 on Build GRCh38
rs747622709 Chr.12: 21765943-21765943 on Build GRCh38
rs866713676 Chr.12: 21765869-21765869 on Build GRCh38
rs556299317 Chr.12: 21765944-21765944 on Build GRCh38
rs188570294 Chr.12: 21765929-21765929 on Build GRCh38
rs746995714 Chr.12: 21766001-21766003 on Build GRCh38
rs912055698 Chr.12: 21765842-21765842 on Build GRCh38
rs763595700 Chr.12: 21765840-21765840 on Build GRCh38
rs775411562 Chr.12: 21765837-21765837 on Build GRCh38
rs144854166 Chr.12: 21766014-21766014 on Build GRCh38
rs970311912 Chr.12: 21765859-21765859 on Build GRCh38

View Full Product Details