Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132080109-132080334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs772241306 Chr.X: 132080237-132080237 on Build GRCh38
rs765049826 Chr.X: 132080161-132080161 on Build GRCh38
rs751171946 Chr.X: 132080182-132080182 on Build GRCh38
rs751277974 Chr.X: 132080295-132080295 on Build GRCh38
rs746397187 Chr.X: 132080203-132080203 on Build GRCh38
rs775394120 Chr.X: 132080276-132080276 on Build GRCh38
rs758060449 Chr.X: 132080172-132080172 on Build GRCh38
rs773237595 Chr.X: 132080269-132080269 on Build GRCh38
rs765102731 Chr.X: 132080272-132080272 on Build GRCh38
rs756807121 Chr.X: 132080296-132080296 on Build GRCh38
rs762876232 Chr.X: 132080286-132080286 on Build GRCh38
rs780549301 Chr.X: 132080194-132080194 on Build GRCh38
rs752422978 Chr.X: 132080164-132080164 on Build GRCh38
rs780567113 Chr.X: 132080209-132080209 on Build GRCh38
rs373562264 Chr.X: 132080152-132080152 on Build GRCh38
rs147353122 Chr.X: 132080218-132080218 on Build GRCh38
rs759421278 Chr.X: 132080160-132080160 on Build GRCh38
rs185450880 Chr.X: 132080235-132080235 on Build GRCh38
rs768888522 Chr.X: 132080222-132080222 on Build GRCh38
rs948849723 Chr.X: 132080221-132080221 on Build GRCh38
rs933474749 Chr.X: 132080173-132080173 on Build GRCh38
rs767487972 Chr.X: 132080147-132080147 on Build GRCh38
rs756646876 Chr.X: 132080208-132080208 on Build GRCh38
rs763767834 Chr.X: 132080289-132080289 on Build GRCh38
rs1043225481 Chr.X: 132080262-132080262 on Build GRCh38
rs983526055 Chr.X: 132080179-132080179 on Build GRCh38
rs759476390 Chr.X: 132080270-132080270 on Build GRCh38
rs767082373 Chr.X: 132080298-132080298 on Build GRCh38
rs774701195 Chr.X: 132080224-132080224 on Build GRCh38
rs756834533 Chr.X: 132080185-132080185 on Build GRCh38

View Full Product Details