Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14493630-14493865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
UniGene
Hs.517819
Gene Chromosome Location
Chr.3: 14489111-14602996 on Build GRCh38
Transcripts
NM_001080423.3 XM_011534141.2 XM_011534142.2 XM_011534143.2 XM_011534145.2 XM_011534146.2

SNP Detail

SNP ID Chromosome Location
rs779700342 Chr.3: 14493652-14493652 on Build GRCh38
rs758050575 Chr.3: 14493648-14493648 on Build GRCh38
rs371141235 Chr.3: 14493764-14493764 on Build GRCh38
rs769147412 Chr.3: 14493700-14493700 on Build GRCh38
rs765627113 Chr.3: 14493808-14493808 on Build GRCh38
rs367805938 Chr.3: 14493786-14493786 on Build GRCh38
rs371618883 Chr.3: 14493800-14493800 on Build GRCh38
rs763154061 Chr.3: 14493679-14493679 on Build GRCh38
rs781148651 Chr.3: 14493658-14493658 on Build GRCh38
rs200524921 Chr.3: 14493675-14493675 on Build GRCh38
rs565224297 Chr.3: 14493723-14493723 on Build GRCh38
rs764418640 Chr.3: 14493737-14493737 on Build GRCh38
rs766983147 Chr.3: 14493815-14493815 on Build GRCh38
rs944336595 Chr.3: 14493714-14493714 on Build GRCh38
rs181971811 Chr.3: 14493699-14493699 on Build GRCh38
rs376935133 Chr.3: 14493813-14493813 on Build GRCh38
rs954854025 Chr.3: 14493781-14493781 on Build GRCh38
rs565606434 Chr.3: 14493722-14493722 on Build GRCh38
rs757111728 Chr.3: 14493844-14493844 on Build GRCh38
rs781001244 Chr.3: 14493721-14493721 on Build GRCh38
rs772124501 Chr.3: 14493735-14493735 on Build GRCh38
rs772103678 Chr.3: 14493798-14493798 on Build GRCh38
rs751150105 Chr.3: 14493715-14493715 on Build GRCh38
rs754226297 Chr.3: 14493738-14493738 on Build GRCh38
rs753190956 Chr.3: 14493700-14493700 on Build GRCh38
rs201727899 Chr.3: 14493759-14493759 on Build GRCh38
rs749111870 Chr.3: 14493773-14493773 on Build GRCh38
rs768451044 Chr.3: 14493657-14493657 on Build GRCh38
rs777232609 Chr.3: 14493802-14493802 on Build GRCh38
rs776313859 Chr.3: 14493805-14493805 on Build GRCh38
rs780461863 Chr.3: 14493692-14493693 on Build GRCh38
rs370347208 Chr.3: 14493676-14493676 on Build GRCh38
rs375473627 Chr.3: 14493769-14493769 on Build GRCh38
rs762233050 Chr.3: 14493746-14493746 on Build GRCh38
rs747473953 Chr.3: 14493699-14493700 on Build GRCh38
rs746099010 Chr.3: 14493791-14493791 on Build GRCh38
rs543809953 Chr.3: 14493799-14493799 on Build GRCh38
rs79010943 Chr.3: 14493817-14493817 on Build GRCh38
rs115041468 Chr.3: 14493833-14493833 on Build GRCh38
rs774516436 Chr.3: 14493731-14493731 on Build GRCh38
rs767939977 Chr.3: 14493696-14493696 on Build GRCh38
rs753857586 Chr.3: 14493807-14493807 on Build GRCh38
rs771111616 Chr.3: 14493729-14493729 on Build GRCh38
rs1006120742 Chr.3: 14493755-14493755 on Build GRCh38
rs909136767 Chr.3: 14493747-14493747 on Build GRCh38
rs747914813 Chr.3: 14493666-14493666 on Build GRCh38
rs964748574 Chr.3: 14493649-14493649 on Build GRCh38
rs759209816 Chr.3: 14493814-14493814 on Build GRCh38
rs375963902 Chr.3: 14493695-14493695 on Build GRCh38
rs35231838 Chr.3: 14493685-14493686 on Build GRCh38
rs756997237 Chr.3: 14493779-14493779 on Build GRCh38
rs764040510 Chr.3: 14493828-14493828 on Build GRCh38
rs766372677 Chr.3: 14493757-14493757 on Build GRCh38
rs372925368 Chr.3: 14493682-14493682 on Build GRCh38
rs370859345 Chr.3: 14493708-14493708 on Build GRCh38
rs775930482 Chr.3: 14493736-14493736 on Build GRCh38
rs749559044 Chr.3: 14493728-14493728 on Build GRCh38
rs148393805 Chr.3: 14493709-14493709 on Build GRCh38
rs950681584 Chr.3: 14493752-14493752 on Build GRCh38
rs867919628 Chr.3: 14493804-14493804 on Build GRCh38
rs767047626 Chr.3: 14493765-14493765 on Build GRCh38
rs759549359 Chr.3: 14493734-14493734 on Build GRCh38
rs373836618 Chr.3: 14493653-14493653 on Build GRCh38
rs546708422 Chr.3: 14493822-14493822 on Build GRCh38
rs747702732 Chr.3: 14493828-14493831 on Build GRCh38
rs768655027 Chr.3: 14493716-14493716 on Build GRCh38
rs376232136 Chr.3: 14493672-14493672 on Build GRCh38
rs777582626 Chr.3: 14493772-14493772 on Build GRCh38
rs773424280 Chr.3: 14493678-14493678 on Build GRCh38
rs756054378 Chr.3: 14493770-14493770 on Build GRCh38
rs923358690 Chr.3: 14493775-14493775 on Build GRCh38

View Full Product Details