Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57092758-57093003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAB2
Entrez Gene ID
4665
Gene Name
NGFI-A binding protein 2
Gene Aliases
MADER
UniGene
Hs.159223
Gene Chromosome Location
Chr.12: 57088894-57095476 on Build GRCh38
Transcripts
NM_005967.3 XM_005268894.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5739694 c.1133T>C p.L378P Chr.12: 57092958-57092958 on Build GRCh38
7211565 c.1143+4T>C p.? Chr.12: 57092972-57092972 on Build GRCh38
5732811 c.1106A>T p.E369V Chr.12: 57092931-57092931 on Build GRCh38
941896 c.1132C>A p.L378M Chr.12: 57092957-57092957 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764973585 Chr.12: 57092888-57092888 on Build GRCh38
rs201943990 Chr.12: 57092871-57092871 on Build GRCh38
rs771321296 Chr.12: 57092957-57092957 on Build GRCh38
rs576129205 Chr.12: 57092804-57092804 on Build GRCh38
rs774634331 Chr.12: 57092968-57092968 on Build GRCh38
rs1033979806 Chr.12: 57092816-57092816 on Build GRCh38
rs919204053 Chr.12: 57092801-57092801 on Build GRCh38
rs324019 Chr.12: 57092864-57092864 on Build GRCh38
rs776133825 Chr.12: 57092881-57092881 on Build GRCh38
rs1043298964 Chr.12: 57092908-57092908 on Build GRCh38
rs749512719 Chr.12: 57092955-57092955 on Build GRCh38
rs376870903 Chr.12: 57092863-57092863 on Build GRCh38
rs746318609 Chr.12: 57092979-57092979 on Build GRCh38
rs572142265 Chr.12: 57092811-57092811 on Build GRCh38
rs781352078 Chr.12: 57092909-57092909 on Build GRCh38
rs973290354 Chr.12: 57092794-57092794 on Build GRCh38
rs376332427 Chr.12: 57092875-57092875 on Build GRCh38
rs748265720 Chr.12: 57092912-57092912 on Build GRCh38
rs912624740 Chr.12: 57092837-57092837 on Build GRCh38
rs992729214 Chr.12: 57092862-57092862 on Build GRCh38
rs199843359 Chr.12: 57092985-57092985 on Build GRCh38
rs376227502 Chr.12: 57092980-57092980 on Build GRCh38
rs181509771 Chr.12: 57092789-57092789 on Build GRCh38
rs201366928 Chr.12: 57092891-57092891 on Build GRCh38
rs756237309 Chr.12: 57092923-57092923 on Build GRCh38
rs35131216 Chr.12: 57092934-57092935 on Build GRCh38
rs941911509 Chr.12: 57092783-57092783 on Build GRCh38
rs958621296 Chr.12: 57092841-57092841 on Build GRCh38
rs766131080 Chr.12: 57092893-57092893 on Build GRCh38
rs528936501 Chr.12: 57092828-57092829 on Build GRCh38
rs761389540 Chr.12: 57092884-57092884 on Build GRCh38
rs760099902 Chr.12: 57092878-57092878 on Build GRCh38
rs763582798 Chr.12: 57092879-57092879 on Build GRCh38
rs751540617 Chr.12: 57092904-57092904 on Build GRCh38
rs749685124 Chr.12: 57092868-57092868 on Build GRCh38
rs755110809 Chr.12: 57092904-57092904 on Build GRCh38
rs773404678 Chr.12: 57092867-57092867 on Build GRCh38
rs373833438 Chr.12: 57092892-57092892 on Build GRCh38
rs561187353 Chr.12: 57092787-57092787 on Build GRCh38
rs778034252 Chr.12: 57092940-57092940 on Build GRCh38
rs558542241 Chr.12: 57092854-57092854 on Build GRCh38
rs1014608966 Chr.12: 57092782-57092782 on Build GRCh38
rs958189063 Chr.12: 57092953-57092953 on Build GRCh38

View Full Product Details